Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
We offer a wide range of detergents and cleaners for clean room applications, medical and clinical disinfection, food and beverage locations, or just general laboratory cleaning. In addition, we offer all the supplies necessary to safely clean up oils, acids, caustics, solvents, and toxic spills.
Decon | Pyroneg | RBS | Virkon S | |
---|---|---|---|---|
Applications | Laboratory, medical, industrial/general | Food, research, industrial, laboratory/general | Food, research, industrial, laboratory/general, proteomics, pharmaceutical, tissue culture | Research, industrial, laboratory/general, proteomics, pharmaceutical |
Process | Manual washing, soaking, ultrasonic cleaners, automatic washers | Manual washing, soaking, ultrasonic cleaners, automatic washers | Manual washing, soaking, ultrasonic cleaners, automatic washers | Manual washing, soaking |
Use | Cleaning/degreasing | Cleaning/degreasing | Cleaning/degreasing | Sterilisation |
Format | Liquid concentrate | Powder or liquid concentrate | Powder or liquid concentrate | Powder |
Non-foaming | ||||
Phosphate free |
Receive a Certificate of Compliance and Certificate(s) of Irradiation (if applicable) on purchased Thermo Scientific Nalgene and Nunc products.
Need technical support? Contact our expert team for technical and application support of Laboratory Products at technicalsupport@thermofisher.com.
For Laboratory Use.