Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Nalgene high density polyethylene bottles are our most popular lab bottles by virtue of their leakproof* performance, compatibility with a wide range of lab reagents, impact resistance down to -100°C, and lower cost than many other materials. High density polyethylene bottles provide very good chemical compatibility with a wide variety of lab reagents, including most acids, bases, alcohols, aldehydes, esters, and aliphatic hydrocarbons.
For chemical compatibility ratings by specific chemical and application temperature, consult the Nalgene Chemical Resistance Guide, or request a copy of the printed wall chart. High density polyethylene bottles are made only from high quality laboratory grade resin materials that comply with a number of regulatory specifications including: USP Class VI, EP monographs, EU food-contact directives, CONEG, RoHS, CA Prop 65, SARA Title III Sec. 313, and 21 CFR pt 177.1520 for food contact. For regulatory claims and certifications applicable to specific Nalgene bottles, please contact Nalgene Regulatory Support.
For help selecting the Thermo Scientific Nalgene bottle that's perfect for your lab applications, use our online selection guide or for personal assistance, contact the Nalgene technical support team.
Looking for ways to make your lab more sustainable? Nalgene high-density polyethylene bottles are reusable, durable and readily recyclable at end-of-life; discover more about Nalgene bottle sustainability and green manufacturing practices through our participation in My Green Lab's ACT Label program.
Narrow-Mouth Round Lab Quality Bottle (2002-series) | Narrow-Mouth Round Economy Bottle (2089-series) | Narrow-Mouth Round Economy Bottle Bulk Pack (332089-series) | Narrow-Mouth Round Amber Lab Quality Bottle (2004-series) | Narrow-Mouth Round Amber Economy Bottle (DS2085-series) |
Traditional reusable lab bottle for long-term use, critical applications | Lower cost, lighter weight, thinner wall for light-duty applications | Bulk packed for additional cost savings, lighter weight, thinner wall for light-duty applications | Amber color protects light-sensitive solutions, traditional reusable lab bottle for long-term use, critical applications | Amber color protects light-sensitive solutions, lower cost, lighter weight, thinner wall for light-duty applications |
![]() | ![]() | ![]() | ![]() | ![]() |
HDPE bottle PP Closure | HDPE bottle PP Closure | HDPE bottle PP Closure | Amber HDPE bottle amber PP closure | Amber HDPE bottle amber PP closure |
Milky white translucent | Milky white translucent | Milky white translucent | Opaque amber bottle | Opaque amber bottle |
4mL–1L | 30mL–1L | 125mL–1L | 4mL–1L | 30mL–1L |
Leakproof* | Leakproof* | Leakproof* | Leakproof* | Leakproof* |
Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC |
Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable |
Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 |
BPA-Free | BPA-Free | BPA-Free | BPA-Free | BPA-Free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
ACT Label | ACT Label | ACT Label | ||
Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled |
Narrow Mouth Round Lab Quality Bottle (2002-series) | Narrow Mouth Round Economy Bottle (2089-series) | Narrow Mouth Round Economy Bottle Bulk Pack (332089-series) | Narrow Mouth Round Amber Lab Quality Bottle (2004-series) | Narrow Mouth Round Amber Economy Bottle (DS2085-series) | |
Acids, dilute or weak | E | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G | G |
Alcohols, aliphatic | E | E | E | E | E |
Aldehydes | G | G | G | G | G |
Bases/alkali | E | E | E | E | E |
Esters | G | G | G | G | G |
Hydrocarbons, aliphatic | G | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N | N |
Ketones, aromatic | N | N | N | N | N |
Oxidizing agents, strong | F | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC |
G | Little or no damage after 30 days constant exposure at 20oC |
F | Some effect after 7 days constant exposure at 20oC |
N | No recommended. Immediate damage may occur |
Wide Mouth Round Lab Quality Bottle (2104-series) | Large Wide Mouth Round Bottle (2120-series) | Wide Mouth Round Economy Bottle (2189-series) | Wide Mouth Round Economy Bottle Bulk Pack (332189-series) | Wide Mouth Round Amber Lab Quality Bottle (2106-series) | Wide Mouth Round Amber Economy Bottle (DS2185-series) |
Traditional reusable lab bottle for long-term use, critical applications | Larger size, rugged lab quality construction | Lower cost, lighter weight, thinner wall for light-duty applications | Bulk packed for additional cost savings, lighter weight, thinner wall for light-duty applications | Amber color protects light-sensitive solutions, traditional reusable lab bottle for long-term use, critical applications | Amber color protects light-sensitive solutions, lower cost, lighter weight, thinner wall for light-duty applications |
![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
HDPE bottle PP Closure | HDPE bottle white PP Closure | HDPE bottle PP Closure | HDPE bottle PP Closure | Amber HDPE bottle amber PP closure | Amber HDPE bottle amber PP closure |
Milky white translucent | Milky white translucent | Milky white translucent | Milky white translucent | Opaque amber bottle | Opaque amber bottle |
30mL–1.5L | 2L–4L | 30mL–1L | 60mL–1L | 30mL–1L | 30mL–1L |
Leakproof* | Not liquid leakproof | Leakproof* | Leakproof* | Leakproof* | Leakproof* |
Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC |
Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable |
Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 |
BPA-Free | BPA-Free | BPA-Free | BPA-Free | BPA-Free | BPA-Free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
ACT Label | ACT Label | ACT Label | |||
Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled |
Wide-Mouth Round Lab Quality Bottle (2104-series) | Large Wide-Mouth Round Bottle (2120-series) | Wide-Mouth Round Economy Bottle (2189-series) | Wide-Mouth Round Economy Bottle Bulk Pack (332189-series) | Wide-Mouth Round Amber Lab Quality Bottle (2106-series) | Wide-Mouth Round Amber Economy Bottle (DS2185-series) | |
Acids, dilute or weak | E | E | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G | G | G |
Alcohols, aliphatic | E | E | E | E | E | E |
Aldehydes | G | G | G | G | G | G |
Bases/alkali | E | E | E | E | E | E |
Esters | G | G | G | G | G | G |
Hydrocarbons, aliphatic | G | G | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N | N | N |
Ketones, aromatic | N | N | N | N | N | N |
Oxidizing agents, strong | F | F | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC. |
G | Little or no damage after 30 days constant exposure at 20oC. |
F | Some effect after 7 days constant exposure at 20oC. |
N | Not recommended. Immediate damage may occur. |
Narrow-Mouth Square HDPE Bottles (2018-series) | Wide-Mouth Square HDPE Bottles (2114-series) | Large Wide-Mouth Square HDPE Bottle (2123-series) |
Space-saving square shape, narrow mouth, graduated | Space-saving square shape, wide mouth | Space-saving square shape, wide mouth, large capacity, molded hand grip |
![]() | ![]() | ![]() |
Milky white translucent | Milky white translucent | Milky white translucent |
30mL–1L | 60mL–1L | 4L |
Leakproof* | Leakproof* | Not liquid leakproof |
Molded graduations | No graduations | No graduations |
Narrow mouth | Wide mouth | Wide mouth |
Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC |
Not autoclavable | Not autoclavable | Not autoclavable |
Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 |
BPA-Free | BPA-Free | BPA-Free |
Phthalate-free | Phthalate-free | Phthalate-free |
ACT Label | ACT Label |
Narrow-Mouth Square HDPE Bottles (2018-series) | Wide-Mouth Square HDPE Bottles (2114-series) | Large Wide-Mouth Square HDPE Bottle (2123-series) | |
Acids, dilute or weak | E | E | E |
Acids^ strong, concentrated | G | G | G |
Alcohols, aliphatic | E | E | E |
Aldehydes | G | G | G |
Bases/alkali | E | E | E |
Esters | G | G | G |
Hydrocarbons, aliphatic | G | G | G |
Hydrocarbons, aromatic | N | N | N |
Hydrocarbons, halogenated | N | N | N |
Ketones, aromatic | N | N | N |
Oxidizing agents, strong | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC |
G | Little or no damage after 30 days constant exposure at 20oC |
F | Some effect after 7 days constant exposure at 20oC |
N | Not recommended. Immediate damage may occur |
Wide-Mouth Rectangular Bottle (2007-series) | Wide-Mouth Amber Rectangular Bottle (2009-series) |
Space-saving rectangular shape | Amber color protects light-sensitive solutions, space-saving rectangular shape |
![]() | ![]() |
Amber HDPE bottle/PP Closure | Amber HDPE bottle/PP Closure |
Milky white translucent | Opaque amber bottle |
125mL–2L | 125mL–2L |
Leakproof* | Leakproof* |
No graduations | No graduations |
Wide mouth | Wide mouth |
Temp range -100oC to 120oC | Temp range -100oC to 120oC |
Not autoclavable | Not autoclavable |
Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 |
BPA-Free | BPA-Free |
Phthalate-free | Phthalate-free |
ACT Label |
Wide-Mouth Rectangular Bottle (2007-series) | Wide-Mouth Amber Rectangular Bottle (2009-series) | |
Acids, dilute or weak | E | E |
Acids^ strong, concentrated | G | G |
Alcohols, aliphatic | E | E |
Aldehydes | G | G |
Bases/alkali | E | E |
Esters | G | G |
Hydrocarbons, aliphatic | G | G |
Hydrocarbons, aromatic | N | N |
Hydrocarbons, halogenated | N | N |
Ketones, aromatic | N | N |
Oxidizing agents, strong | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC |
G | Little or no damage after 30 days constant exposure at 20oC |
F | Some effect after 7 days constant exposure at 20oC |
N | Not recommended. Immediate damage may occur |
Narrow-Mouth Round Lab Quality Bottle (2002-series) | Narrow-Mouth Round Economy Bottle (2089-series) | Narrow-Mouth Round Economy Bottle Bulk Pack (332089-series) | Narrow-Mouth Round Amber Lab Quality Bottle (2004-series) | Narrow-Mouth Round Amber Economy Bottle (DS2085-series) |
Traditional reusable lab bottle for long-term use, critical applications | Lower cost, lighter weight, thinner wall for light-duty applications | Bulk packed for additional cost savings, lighter weight, thinner wall for light-duty applications | Amber color protects light-sensitive solutions, traditional reusable lab bottle for long-term use, critical applications | Amber color protects light-sensitive solutions, lower cost, lighter weight, thinner wall for light-duty applications |
![]() | ![]() | ![]() | ![]() | ![]() |
HDPE bottle PP Closure | HDPE bottle PP Closure | HDPE bottle PP Closure | Amber HDPE bottle amber PP closure | Amber HDPE bottle amber PP closure |
Milky white translucent | Milky white translucent | Milky white translucent | Opaque amber bottle | Opaque amber bottle |
4mL–1L | 30mL–1L | 125mL–1L | 4mL–1L | 30mL–1L |
Leakproof* | Leakproof* | Leakproof* | Leakproof* | Leakproof* |
Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC |
Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable |
Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 |
BPA-Free | BPA-Free | BPA-Free | BPA-Free | BPA-Free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
ACT Label | ACT Label | ACT Label | ||
Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled |
Narrow Mouth Round Lab Quality Bottle (2002-series) | Narrow Mouth Round Economy Bottle (2089-series) | Narrow Mouth Round Economy Bottle Bulk Pack (332089-series) | Narrow Mouth Round Amber Lab Quality Bottle (2004-series) | Narrow Mouth Round Amber Economy Bottle (DS2085-series) | |
Acids, dilute or weak | E | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G | G |
Alcohols, aliphatic | E | E | E | E | E |
Aldehydes | G | G | G | G | G |
Bases/alkali | E | E | E | E | E |
Esters | G | G | G | G | G |
Hydrocarbons, aliphatic | G | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N | N |
Ketones, aromatic | N | N | N | N | N |
Oxidizing agents, strong | F | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC |
G | Little or no damage after 30 days constant exposure at 20oC |
F | Some effect after 7 days constant exposure at 20oC |
N | No recommended. Immediate damage may occur |
Wide Mouth Round Lab Quality Bottle (2104-series) | Large Wide Mouth Round Bottle (2120-series) | Wide Mouth Round Economy Bottle (2189-series) | Wide Mouth Round Economy Bottle Bulk Pack (332189-series) | Wide Mouth Round Amber Lab Quality Bottle (2106-series) | Wide Mouth Round Amber Economy Bottle (DS2185-series) |
Traditional reusable lab bottle for long-term use, critical applications | Larger size, rugged lab quality construction | Lower cost, lighter weight, thinner wall for light-duty applications | Bulk packed for additional cost savings, lighter weight, thinner wall for light-duty applications | Amber color protects light-sensitive solutions, traditional reusable lab bottle for long-term use, critical applications | Amber color protects light-sensitive solutions, lower cost, lighter weight, thinner wall for light-duty applications |
![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
HDPE bottle PP Closure | HDPE bottle white PP Closure | HDPE bottle PP Closure | HDPE bottle PP Closure | Amber HDPE bottle amber PP closure | Amber HDPE bottle amber PP closure |
Milky white translucent | Milky white translucent | Milky white translucent | Milky white translucent | Opaque amber bottle | Opaque amber bottle |
30mL–1.5L | 2L–4L | 30mL–1L | 60mL–1L | 30mL–1L | 30mL–1L |
Leakproof* | Not liquid leakproof | Leakproof* | Leakproof* | Leakproof* | Leakproof* |
Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC |
Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable |
Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 |
BPA-Free | BPA-Free | BPA-Free | BPA-Free | BPA-Free | BPA-Free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
ACT Label | ACT Label | ACT Label | |||
Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled |
Wide-Mouth Round Lab Quality Bottle (2104-series) | Large Wide-Mouth Round Bottle (2120-series) | Wide-Mouth Round Economy Bottle (2189-series) | Wide-Mouth Round Economy Bottle Bulk Pack (332189-series) | Wide-Mouth Round Amber Lab Quality Bottle (2106-series) | Wide-Mouth Round Amber Economy Bottle (DS2185-series) | |
Acids, dilute or weak | E | E | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G | G | G |
Alcohols, aliphatic | E | E | E | E | E | E |
Aldehydes | G | G | G | G | G | G |
Bases/alkali | E | E | E | E | E | E |
Esters | G | G | G | G | G | G |
Hydrocarbons, aliphatic | G | G | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N | N | N |
Ketones, aromatic | N | N | N | N | N | N |
Oxidizing agents, strong | F | F | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC. |
G | Little or no damage after 30 days constant exposure at 20oC. |
F | Some effect after 7 days constant exposure at 20oC. |
N | Not recommended. Immediate damage may occur. |
Narrow-Mouth Square HDPE Bottles (2018-series) | Wide-Mouth Square HDPE Bottles (2114-series) | Large Wide-Mouth Square HDPE Bottle (2123-series) |
Space-saving square shape, narrow mouth, graduated | Space-saving square shape, wide mouth | Space-saving square shape, wide mouth, large capacity, molded hand grip |
![]() | ![]() | ![]() |
Milky white translucent | Milky white translucent | Milky white translucent |
30mL–1L | 60mL–1L | 4L |
Leakproof* | Leakproof* | Not liquid leakproof |
Molded graduations | No graduations | No graduations |
Narrow mouth | Wide mouth | Wide mouth |
Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC |
Not autoclavable | Not autoclavable | Not autoclavable |
Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 |
BPA-Free | BPA-Free | BPA-Free |
Phthalate-free | Phthalate-free | Phthalate-free |
ACT Label | ACT Label |
Narrow-Mouth Square HDPE Bottles (2018-series) | Wide-Mouth Square HDPE Bottles (2114-series) | Large Wide-Mouth Square HDPE Bottle (2123-series) | |
Acids, dilute or weak | E | E | E |
Acids^ strong, concentrated | G | G | G |
Alcohols, aliphatic | E | E | E |
Aldehydes | G | G | G |
Bases/alkali | E | E | E |
Esters | G | G | G |
Hydrocarbons, aliphatic | G | G | G |
Hydrocarbons, aromatic | N | N | N |
Hydrocarbons, halogenated | N | N | N |
Ketones, aromatic | N | N | N |
Oxidizing agents, strong | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC |
G | Little or no damage after 30 days constant exposure at 20oC |
F | Some effect after 7 days constant exposure at 20oC |
N | Not recommended. Immediate damage may occur |
Wide-Mouth Rectangular Bottle (2007-series) | Wide-Mouth Amber Rectangular Bottle (2009-series) |
Space-saving rectangular shape | Amber color protects light-sensitive solutions, space-saving rectangular shape |
![]() | ![]() |
Amber HDPE bottle/PP Closure | Amber HDPE bottle/PP Closure |
Milky white translucent | Opaque amber bottle |
125mL–2L | 125mL–2L |
Leakproof* | Leakproof* |
No graduations | No graduations |
Wide mouth | Wide mouth |
Temp range -100oC to 120oC | Temp range -100oC to 120oC |
Not autoclavable | Not autoclavable |
Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 |
BPA-Free | BPA-Free |
Phthalate-free | Phthalate-free |
ACT Label |
Wide-Mouth Rectangular Bottle (2007-series) | Wide-Mouth Amber Rectangular Bottle (2009-series) | |
Acids, dilute or weak | E | E |
Acids^ strong, concentrated | G | G |
Alcohols, aliphatic | E | E |
Aldehydes | G | G |
Bases/alkali | E | E |
Esters | G | G |
Hydrocarbons, aliphatic | G | G |
Hydrocarbons, aromatic | N | N |
Hydrocarbons, halogenated | N | N |
Ketones, aromatic | N | N |
Oxidizing agents, strong | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC |
G | Little or no damage after 30 days constant exposure at 20oC |
F | Some effect after 7 days constant exposure at 20oC |
N | Not recommended. Immediate damage may occur |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
These HDPE bottles are manufactured in a controlled environment in order to minimise the risk of contamination with raw materials specially selected to meet the rigorous requirements demanded by analytical laboratories for cleanliness and purity.
When looking for sustainable products, we believe it should be easy to make informed purchasing decisions. We're working to provide more information about our products' environmental impact by participating in the ACT Label program with a selection of Thermo Scientific Nalgene and Matrix products. It's like an eco-nutrition label for sustainability.