Search Thermo Fisher Scientific
Johne's disease, also known as paratuberculosis, is a global health issue affecting ruminants and is a result of an infection with Mycobacterium avium spp. paratuberculosis (MAP). Primarily affecting cattle, sheep, and goats, this disease can lead to severe production constraints and substantial economic losses within livestock populations due to its lengthy incubation period, which makes diagnosis challenging. By offering accurate and reliable diagnostic solutions, we aim to assist producers, veterinarians, and researchers in effectively managing and controlling Johne's Disease outbreaks.
The key to controlling the disease lies in the early detection of subclinical disease in animals. These animals can excrete the organism over extended periods, posing an infection risk to the rest of the herd. Utilizing a combination of diagnostic methods such as ELISA and real-time PCR tests can aid in identifying and isolating these "shedders" at an earlier stage, thereby mitigating the risk of infection to other healthy animals in the herd.
Our diagnostic solutions offer reliable and accurate results, providing an essential tool in the battle against paratuberculosis.
The PrioCHECK MAP Ab 3.0 Plate Kit is an updated, easy-to-use ELISA kit for the detection of antibodies against MAP/paratuberculosis which cause Johne's Disease. Optimized for high performance and efficiency in the lab, the improved kit is now operating on a universal protocol across compatible species and sample types. This means no more juggling different protocols for different species; our kit simplifies your workflow while maintaining high precision.
|
Species |
||
Sample type |
Cattle |
Sheep |
Goat |
|
|
|
Our PrioCHECK Paratuberculosis Ab 3.0 Plate Kit is engineered to streamline your laboratory work with a host of practical features:
The PARACHECK 2 Kit is a solid phase, indirect enzyme immunoassay (EIA) to detect antibodies to M. paratuberculosis in animal sera and milk of cattle, sheep, and goats. The PARACHECK 2 procedure for 96 determinations (including controls) takes approximately 80 min and consists of four steps. In the first step, the samples are diluted and incubated in a diluent buffer containing M. phlei to remove cross-reacting antibodies. In the second step, specific antibodies present in the diluted samples bind to M. paratuberculosis antigens coated to the wells of a microtiter plate. Unreacted proteins are removed by washing. A secondary antibody that is conjugated to an enzyme that generates a color signal is added in the third step. In the fourth step, the presence of specific antibodies is visualized by adding the enzyme substrate. The intensity of the color signal is proportional to the amount of antibodies in the sample.
Real-time PCR enables sensitive and specific detection of pathogen nucleic acid in animal samples, allowing for reliable and rapid screening and detection of infected animals.
The VetMAX M. paratuberculosis 2.0 Kit kits include:
How has Mycobacterium avium ssp. paratuberculosis (MAP) been monitored in Saxony, Germany and how did real-time PCR perform in field testing? Presentation of Dr. Christina Boss, senior product manager of Dairy Solutions at Thermo Fisher Scientific, discusses the findings in the presentation of Dr. Andrea Konrath, Health and Veterinary Research Institute of Saxony, given at the Bacteriology AVID-Tagung in September 2015.
The VetMAX real-time PCR tests are part of a fully integrated, single-vendor workflow that includes the Applied Biosystems MagMAX system for sample preparation and extraction and the Applied Biosystems 7500 Real-Time PCR System.
For Veterinary Use Only. For In Vitro Use Only. Regulatory requirements vary by country; products may not be available in your geographic area.