Search Thermo Fisher Scientific
Applied Biosystems MicroSEQ Microbial Identification Software is part of the integrated workflow for the identification and classification of bacterial and fungal sequences for scientists working in a QC microbiology laboratory setting. The software is used to collect, analyze, and compare sequence data to the species level.
Identify and classify your bacterial or fungal sequence using the latest, feature-rich release of our tried-and-true analysis software, specifically designed for the Applied Biosystems SeqStudio family of genetic analyzers. MicroSEQ Identification Software 4.0 allows users to leverage the advanced features of the SeqStudio systems while also helping meet regulatory requirements to enable 21 CFR Part 11 compliance.
One of the keys to accurate microbial identification, and the engine driving MicroSEQ ID, is a robust and powerful database. We don't just collect data from other sources. We validate our own findings and confirm accuracy of all entries. The MicroSEQ ID System is built on meticulously curated collections of organisms—more than 12,000 strains—validated by our own scientists and collaborators using multiple techniques to enable accurate identification
Thermo Fisher Scientific conducts a rigorous validation process for accurate taxonomic information of new and existing bacterial and fungal species. From sequence information entry, genus and species level validation, to revalidation of existing MicroSEQ library entries—all done by trained personnel—you are offered one of the most comprehensive, accurate, reliable libraries available for your Pharma QC testing.
Curated:
We closely evaluate our libraries and curate for what really matters to our pharma QC customers—quality and accuracy. We have increased the number of Candida, Cladosproium, Aspergillus, and added species for 223 new genera into our fungal database. We partner with the Westerdijk Fungal Biodiversity Institute, part of the Royal Netherlands Academy of Arts and Sciences, for the latest and most reliable sequence information for mycological identification and classification.
Increasing the power of this database, Thermo Fisher Scientific regularly updates the MicroSEQ ID Software Library. This helps ensure that your MicroSEQ ID database is always up to date by providing periodic updates of quality checked and validated bacterial and fungal sequences.
Simple user interface and navigation with “Get Started” buttons and a left navigation panel.
Easy-to-read run results. The status of each well appears in real-time. Report links are provided on the right side of the screen.
Easy to analyze and review the results in various report formats and color-coded quality report out.
Simple user interface and navigation with “Get Started” buttons and a left navigation panel.
Easy-to-read run results. The status of each well appears in real-time. Report links are provided on the right side of the screen.
Easy to analyze and review the results in various report formats and color-coded quality report out.
v3.1.3 | v1.0 | v4.0 | |
Instrument support | 3500 / 3500 XL | SeqStudio Genetic Analyzer | SeqStudio and SeqStudio Flex Systems |
MSID Kits compatible | Yes | Yes | Yes |
MSID Libraries compatible | Yes | Yes | Yes |
Compatible with SAE Administration Console | No | No | Yes |
Multiple concurrent | No | No | Yes |
SAE compliance | Yes | Yes | Yes, with enhanced, single SAE for instrument & SW |
Technology | Desktop app | Desktop app | Browser-based |
Data archive / restore | Audit only | Audit only | Archive and backup / restore |
Advanced reporting | Sample & project reporting | Sample & project reporting | Sample & project reporting with customizable output |
Explore SeqStudio QST and SeqStudio Flex PA Genetic Analyzers >
Learn more about the MicroSEQ ID genotypic approach >
Explore the Analytics Knowledge Hub from Thermo Fisher Scientific, providing a robust collection of webinars, articles, eBooks, infographics, and other digital resources to help you optimize the use of our products in your bioprocess workflow.
Explore the Hub