Search Thermo Fisher Scientific
Zinc concentrations in the 1–100 nM range are routinely measured using fluorescent indicators nominally designed for Ca2+ detection, but higher Zn2+ concentrations are present in synaptic vesicles and released in response to electrical stimulation or excitotoxic agonists.
We’ve developed a number of Molecular Probes ion indicators to track zinc with intense fluorescent signals and a range of wavelength options.
FluoZin dyes are unique indicators designed for detection of Zn2+ in the 0.1–100 µM range with minimal interfering Ca2+ sensitivity. They provide bright signals for assaying exocytotic release of Zn2+ from pancreatic β-cells after stimulation with glucose or potassium. Zn2+ concentrations can be measured using imaging platforms, cell-based microplate assays, or flow cytometry.
Used for spatial and temporal resolution of Zn2+ concentrations.
FluoZin-1, Tripotassium Salt | FluoZin-1, AM | FluoZin-3, Tetrapotassium Salt | FluoZin-3, AM | |
---|---|---|---|---|
Readout | Responds to Zn2+ concentrations in synaptic vesicles with increasing fluorescence | Responds to saturating levels of Zn2+ with 50-fold increase in fluorescence emission | ||
Range | 0–100 µM range with minimal interfering Ca2+ sensitivity | Low nM to µM range with minimal interfering Ca2+ sensitivity | ||
Fluorophore | FluoZin-1 | FluoZin-3 | ||
Standard filter set | FITC | |||
Ex/Em (nm) | 495/515 | 494/516 | ||
Bibliography | Citations | Citations | ||
Cell permeant or impermeant? | Impermeant | Permeant | Impermeant | Permeant |
Platforms | Imaging | Imaging | Imaging | Imaging |
Usage notes | Load by microinjection, patch pipette, or pinocytic loading agent | Load in cell media | Load by microinjection, patch pipette, or pinocytic loading agent | Load in cell media |
Format | 500 µg | 50 µg | 500 µg | 100 µg |
Cat. No. | F24180 | F24181 | F24194 | F24195 |