Search Thermo Fisher Scientific
MicroAmp 8-Tube Strips, Cap Strips, & Individual PCR Tubes |
PCR tubes are laboratory staples that are critical to the success of an experiment. Applied Biosystems MicroAmp tubes, MicroAmp 8-Tube Strips, and MicroAmp Cap Strips are designed for precision and accuracy in PCR and qPCR experiments.
These single tubes are suitable for small scale experiments (1–3 reactions). Some of our popular MicroAmp PCR tubes are listed in Table 1.
Product | MicroAmp Fast 8-Tube Strip, 0.1 mL | MicroAmp Optical 8-Tube Strip, 0.2 mL | MicroAmp Optical 8-Cap Strips | MicroAmp Optical 8-Tube Strip with Attached Optical Caps, 0.2 mL | MicroAmp 8-Tube Strip with Attached Domed Caps, 0.2 mL |
Cap type | Not included | Not included | Optical | Optical | Domed |
Applied Biosystems MicroAmp 8-Tube Strip with attached optical or domed caps offers a combination of features to help prevent cross-contamination, pipetting errors, and sample identification errors.
The PCR plastics Sustain Series are ISCC PLUS-certified, bio-based PCR plastics that help reduce lab’s carbon footprint. This represents our commitment to integrating more sustainable practices into our manufacturing processes, without compromising on product quality or performance and are certified free of DNases, RNases, human DNA, and PCR inhibitors to help ensure the accuracy and reproducibility of PCR results.
PCR enzymes education resources | |
Access PCR learning resources—a knowledge base of all things PCR. |
Molecular biology webinars | |
View our webinars for trends, tips, and tricks on molecular biology techniques. |
PCR plastics videos | |
Watch videos on how to select and use the right consumables for your PCR and qPCR experiments. |
Support center | |
Find tips, troubleshooting help, and resources for your PCR and cDNA synthesis experiments. |
Contact us | |
Email or call our Technical Application Scientists for questions regarding products for PCR workflow. |
OEM and custom PCR products | |
Discover custom manufacturing solutions, product barcoding, and packaging capabilities tailored to your specifications. |