Search Thermo Fisher Scientific
ProtoArray services and its associated products have been discontinued, information here is for reference.
The ProtoArray Human Protein Microarray is a high-density microarray that contains thousands of unique, full-length human proteins, including kinases, phosphatases, GPCRs, nuclear receptors, and proteases, spotted in duplicate on a 1 inch x 3 inch thin nitrocellulose coated glass slide. ProtoArray human protein microarray contains over 9,000 unique human proteins individually purified and arrayed under native conditions to maximize functionality.
The microarray includes control ProtoArray Human Protein Microarray proteins in each sub-array to facilitate supported applications. Additionally, data analysis software, ProtoArray Prospector, is freely available for examining the results from your experiments.