Search Thermo Fisher Scientific
Streamline your western blotting workflow by empowering your lab with the Invitrogen iWestern workflow devices and instruments. Designed to address processing efficiency, reproducibility, and robustness of results, the iWestern workflow helps to enable exceptional western blot results with minimal hands-on time.
The iWestern workflow is a four-step approach to streamline your western blotting protocols, incorporating our high-quality Invitrogen western blot benchtop devices and instruments. Used together, these tools simplify the western workflow to obtain quality results faster than traditional manual western blotting.
Invitrogen PowerEase Touch Power Supplies feature a large touch screen that is easy to operate and program while delivering precise outputs for voltage, current, and power. Ideal for applications like SDS-PAGE, native PAGE, and DNA/RNA electrophoresis.
With an intelligently engineered side-by-side design, the Invitrogen Mini Gel Tank provides forward-facing well configuration for easier sample loading and the simultaneous visualization of both gels, while the white-colored tank stand provides contrast to improve the monitoring of pre-stained markers during electrophoresis.
Invitrogen Surelock Tandem Midi Gel tank is designed for easy and consistent vertical protein gel electrophoresis of one or two higher-throughput, wider format Invitrogen midi gels.
The iBlot 3 Western Blot Transfer System is a next-generation dry protein transfer device with ready-to-use consumables that delivers high performance and convenience. The iBlot 3 uses pre-assembled transfer stacks with transfer buffer incorporated into gel matrices, so there is no need to prepare transfer buffers and there is minimal post-transfer cleanup required. The short distance between electrodes, along with high field strength and current, reduces transfer time to as few as three minutes.
Invitrogen Power Blotter Transfer System is a versatile semi-dry transfer system for the performance you need and flexibility you want. The Power Blotter features interchangeable blotting cassettes to suit your required throughput. Use your choice of membranes and filter papers or, for added convenience, use Power Blotter Select Transfer Stacks which eliminate the need to prepare transfer buffer by incorporating it as a gel matrix directly in the transfer stack.
Two devices, one simple technology for western blot processing. The Invitrogen iBind Western Systems are simple, non-powered devices that automate many of the tedious, routine western blot processing steps. The original iBind Western Device accommodates the processing of one mini blot at a time, while the iBind Flex Western Device accommodates the processing of up to one midi blot, two mini blots, or up to six vertically cut strips. The entire immunoblotting process is complete in less than three hours, uses up to 80% less primary antibody than traditional methods, and because processing is automated, blot-to-blot consistency can be improved.
iBind Western System
iBind Flex Western System
Experience hands-free blot processing using your protocols with the Invitrogen Bandmate Automated Western Blot Processor. The Bandmate Automated Western Blot Processor is a programmable blot-rocking system that frees up time by automating the hands-on blocking, washing, and antibody incubation steps of western blot processing. Minimal effort is required to set up the device to process up to four mini blots or two midi blots.
Experience an easier time capturing and analyzing data from gels and western blots with Invitrogen iBright Imaging Systems. Designed with a streamlined, intuitive interface and workflows, iBright Imaging Systems are easy to use for researchers of all experience levels.
There are three models in the iBright Imaging Systems family: the iBright CL750 Imaging System, the iBright CL1500 Imaging System, and the iBright FL1500 Imaging System. The iBright CL750 Imaging System offers the core essential western blot and gel imaging functions and makes the transition from the darkroom and film easy. The iBright CL1500 Imaging System expands application support and has many of the high-performance specifications of our premier iBright FL1500 model. The iBright FL1500 Imaging System features maximum application support, including fluorescent western blot imaging with up to four fluorescence channels at a time.
*Note: automatic rotation available only in iBright FL1500 and iBright CL1500 models.
Build your own customized iWestern bundle or select from our pre-configured smaller bundles for each of our western blot devices with built-in savings to get started.
Contents | Cat. No. | |
---|---|---|
PowerEase Touch 120 W Power Supply and Mini Gel Tank Welcome Pack |
| |
PowerEase Touch 350 W Power Supply and SureLock Tandem Midi Gel Tank Welcome Pack |
| |
Bolt Bis-Tris 4–12%, 10-well Welcome Pack |
| |
NuPAGE Bis-Tris 4–12%, 10-well Welcome Pack |
| |
NuPAGE Tris-Glycine 4–20%, 10-well Welcome Pack |
| |
NuPAGE Tris-Acetate 3–8%, 10-well Welcome Pack |
| |
NuPAGE Midi Bis-Tris Welcome Pack, 4–12%, 20-well |
| |
Novex Midi Tris-Glycine Plus Welcome Pack, 4–20%, 20-well |
|
Contents | Cat. No | |
---|---|---|
iBlot 3 starter Kit |
| IB31001S (nitrocellulose) IB31002S (PVDF) |
iBlot 3 Starter Kit with Warranty |
| A56728 (nitrocellulose) A56729 (PVDF) |
iBlot 3 iBind Starter Kit |
| IB31001SLF1 (nitrocellulose) |
iBlot 3 iBind Flex Starter Kit |
| IB31001SLF2 (nitrocellulose) |
Bolt Welcome Pack with iBlot 3 |
|
Contents | Cat. No | |
---|---|---|
| SLF2002S (fluorescent detection) | |
iBind Western Starter Kit |
| SLF1002S (fluorescent detection) |
The iWestern workflow is a four-step approach to streamline your western blotting protocols, incorporating our high-quality Invitrogen western blot benchtop devices and instruments. Used together, these tools simplify the western workflow to obtain quality results faster than traditional manual western blotting.
Invitrogen PowerEase Touch Power Supplies feature a large touch screen that is easy to operate and program while delivering precise outputs for voltage, current, and power. Ideal for applications like SDS-PAGE, native PAGE, and DNA/RNA electrophoresis.
With an intelligently engineered side-by-side design, the Invitrogen Mini Gel Tank provides forward-facing well configuration for easier sample loading and the simultaneous visualization of both gels, while the white-colored tank stand provides contrast to improve the monitoring of pre-stained markers during electrophoresis.
Invitrogen Surelock Tandem Midi Gel tank is designed for easy and consistent vertical protein gel electrophoresis of one or two higher-throughput, wider format Invitrogen midi gels.
The iBlot 3 Western Blot Transfer System is a next-generation dry protein transfer device with ready-to-use consumables that delivers high performance and convenience. The iBlot 3 uses pre-assembled transfer stacks with transfer buffer incorporated into gel matrices, so there is no need to prepare transfer buffers and there is minimal post-transfer cleanup required. The short distance between electrodes, along with high field strength and current, reduces transfer time to as few as three minutes.
Invitrogen Power Blotter Transfer System is a versatile semi-dry transfer system for the performance you need and flexibility you want. The Power Blotter features interchangeable blotting cassettes to suit your required throughput. Use your choice of membranes and filter papers or, for added convenience, use Power Blotter Select Transfer Stacks which eliminate the need to prepare transfer buffer by incorporating it as a gel matrix directly in the transfer stack.
Two devices, one simple technology for western blot processing. The Invitrogen iBind Western Systems are simple, non-powered devices that automate many of the tedious, routine western blot processing steps. The original iBind Western Device accommodates the processing of one mini blot at a time, while the iBind Flex Western Device accommodates the processing of up to one midi blot, two mini blots, or up to six vertically cut strips. The entire immunoblotting process is complete in less than three hours, uses up to 80% less primary antibody than traditional methods, and because processing is automated, blot-to-blot consistency can be improved.
iBind Western System
iBind Flex Western System
Experience hands-free blot processing using your protocols with the Invitrogen Bandmate Automated Western Blot Processor. The Bandmate Automated Western Blot Processor is a programmable blot-rocking system that frees up time by automating the hands-on blocking, washing, and antibody incubation steps of western blot processing. Minimal effort is required to set up the device to process up to four mini blots or two midi blots.
Experience an easier time capturing and analyzing data from gels and western blots with Invitrogen iBright Imaging Systems. Designed with a streamlined, intuitive interface and workflows, iBright Imaging Systems are easy to use for researchers of all experience levels.
There are three models in the iBright Imaging Systems family: the iBright CL750 Imaging System, the iBright CL1500 Imaging System, and the iBright FL1500 Imaging System. The iBright CL750 Imaging System offers the core essential western blot and gel imaging functions and makes the transition from the darkroom and film easy. The iBright CL1500 Imaging System expands application support and has many of the high-performance specifications of our premier iBright FL1500 model. The iBright FL1500 Imaging System features maximum application support, including fluorescent western blot imaging with up to four fluorescence channels at a time.
*Note: automatic rotation available only in iBright FL1500 and iBright CL1500 models.
Build your own customized iWestern bundle or select from our pre-configured smaller bundles for each of our western blot devices with built-in savings to get started.
Contents | Cat. No. | |
---|---|---|
PowerEase Touch 120 W Power Supply and Mini Gel Tank Welcome Pack |
| |
PowerEase Touch 350 W Power Supply and SureLock Tandem Midi Gel Tank Welcome Pack |
| |
Bolt Bis-Tris 4–12%, 10-well Welcome Pack |
| |
NuPAGE Bis-Tris 4–12%, 10-well Welcome Pack |
| |
NuPAGE Tris-Glycine 4–20%, 10-well Welcome Pack |
| |
NuPAGE Tris-Acetate 3–8%, 10-well Welcome Pack |
| |
NuPAGE Midi Bis-Tris Welcome Pack, 4–12%, 20-well |
| |
Novex Midi Tris-Glycine Plus Welcome Pack, 4–20%, 20-well |
|
Contents | Cat. No | |
---|---|---|
iBlot 3 starter Kit |
| IB31001S (nitrocellulose) IB31002S (PVDF) |
iBlot 3 Starter Kit with Warranty |
| A56728 (nitrocellulose) A56729 (PVDF) |
iBlot 3 iBind Starter Kit |
| IB31001SLF1 (nitrocellulose) |
iBlot 3 iBind Flex Starter Kit |
| IB31001SLF2 (nitrocellulose) |
Bolt Welcome Pack with iBlot 3 |
|
Contents | Cat. No | |
---|---|---|
| SLF2002S (fluorescent detection) | |
iBind Western Starter Kit |
| SLF1002S (fluorescent detection) |