Search Thermo Fisher Scientific
Mammalian transient expression systems enable flexible and rapid production of proteins. They're ideal for expression of human or other mammalian proteins because these systems generate recombinant proteins with more native folding and post-translational modifications—such as glycosylation—than expression systems based on hosts such as E. coli, yeast, or insect cells.
FreeStyle 293 SystemTraditional transient 293 expression system | Expi293 | ExpiCHO | |
---|---|---|---|
Protein yield* | 20–200 mg/L | Up to 1 g/L | Up to 3 g/L |
Cells | FreeStyle 293-F | Expi293-F | ExpiCHO-S |
Media | FreeStyle 293 Medium | Expi293 Medium | ExpiCHO Medium |
Transfection reagent | 293Fectin | ExpiFectamine 293 Transfection Kit | ExpiFectamine CHO Transfection Kit |
Positive control vector | pCMV•Sport-βgal | Antibody-expressing vector | Antibody-expressing vector |
Maximum working culture Density | 2 x 106/mL | 6 x 106/mL | 10 x 106/mL |
Cell density for transfection | 1 x 106/mL | 2.5 x 106/mL | 6 x 106/mL |
Order now | |||
*Typical yield of human IgGs |
NEW | Protein Expression Support Center Find tips, troubleshooting help, and resources for your protein expression applications. |