Search Thermo Fisher Scientific
Prefix | Symbol | Factor |
---|---|---|
exa | E | 1018 |
peta | P | 1015 |
tera | T | 1012 |
giga | G | 109 |
mega | M | 106 |
kilo | k | 103 |
hecto | h | 102 |
deca | da | 101 |
deci | d | 10-1 |
centi | c | 10-2 |
mili | m | 10-3 |
micro | µ | 10-6 |
nano | n | 10-9 |
pico | p | 10-12 |
femto | f | 10-15 |
atto | a | 10-18 |
zepto | z | 10-21 |
quaco* | g | 10-23 |