Search Thermo Fisher Scientific
Organism Type | Dog | Madin-Darby canine kidney Media used: MEM cat# 11095 w/ 10% FBS. ATTC cat# CCL-34 |
Tissues | Kidney | |
Phenotype | Adherent | |
Primary | no | |
Application | Cell Culture / Growth Conditions,Protein Expression,Stable Cell Transfection,Transfection: Gene Expression,Transfection: Gene Silencing,Transient Transfection |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||