Search Thermo Fisher Scientific
华夏™白金PCR扩增试剂盒涵盖了华夏PCR扩增试剂盒的16个STR位点,包含最新的COIDS核心位点,并且综合了中国公安部国家DNA数据库常见的位点和Applied Biosystems™其他试剂盒所包含的位点,新增了8个STR位点。基于成熟的六色荧光化学技术、针对中国人群包括和部分少数民族人群进行重新设计,根据中国使用需求对试剂盒标准提出严格定义。华夏白金PCR扩增试剂盒具备更加出色的使用效果和更加强大的个体识别能力。