Search Thermo Fisher Scientific
环境空气质量自动监测系统可对环境空气质量进行24小时自动连续监测。该系统由监测中心站、监测子站和质量保证实验室组成。其中环境空气监测子站包括采样系统、气体分析仪器、校准装置、气象系统、子站数据采集等。子站监测的数据通过多种通讯方式传送至环境监测中心站进行实时控制、数据管理及图表生成。
VOCs在线监测系统示意图
作为无人值守7天/24小时分析运行的在线VOCs监测系统,其组成至少应包括采样装置、分析仪、多点校准仪、零气发生器、氢气和载气发生器(或钢瓶)、数据采集器以及符合要求的分析站房,而且必须保证系统各部分之间配套性、稳定性等达到最佳性能,以保证系统的长期稳定运行。系统至少应包括仪器控制、自动校准或检查、数据分析处理和存储、监测结果的实时上传等功能。
在线VOCs富集系统和ISQ气质联用仪(GCMS)通过创新系统设计、灵活软硬件配置、优化分析方法可实现对空气中VOCs组分的24小时/7天连续定性定量分析。该方案可以用于在线环境监测以及离线环境监测等多种场合,也可以用于车载,实现环境应急监测。
采用技术成熟、性能稳定的GC-FID技术,加上先进的在线VOCs富集系统设计,采用直接法对环境空气中的甲烷和非甲烷总烃进行24小时x7天连续定量分析。