Search Thermo Fisher Scientific
GeneArt™ Plasmid Services include flexible offerings covering construction of plasmid vectors customized for your needs, and subcloning of your sequence into any vector, including the Gateway® vector system. Concentrate on your research goals and leave your plasmid construction and subcloning work to us.
Experienced scientists, using proven protocols and quality-controlled systems and reagents prepare purified plasmid DNA (pDNA) for GeneArt™ Plasmid Services. Our goal is to provide consistent high quality plasmid preparations for research applications and preclinical studies.
Highly pure, homogeneous plasmid DNA: transfection/transformation-ready
Applications include:
Download the Plasmid Order Form, enter your project using Microsoft Excel, and email it to the GeneArt® Plasmid Services team at GeneArtSupport@lifetech.com. |
When setting up your project online, choose “plasmid preparation” and follow the instructions to enter your plasmid preparation requirements. |
Characteristic | Method | Specification |
---|---|---|
Visual appearance | Visual inspection | Clear, Colorless solution |
Homogeneity | Agarose gel electrophoresis (AGE) | ≥85% super-coiled (dependent on plasmid characteristics) |
Concentration | UV spectrophotometry: A260 | 0.5–2 mg/mL or as specified |
Identity | Restriction digestion, AGE | Comigration with appropriate size standard |
A260/280 ratio | UV spectrophotometry | 1.8–2.0 |
Endotoxin content | LAL assay (optional) | ≤0.1 EU/µg pDNA (<0.01 EU/µg pDNA available) |
For Research Use Only. Not for use in diagnostic procedures.