Search Thermo Fisher Scientific
Insufficient nucleic acid extraction is the most common cause of molecular workflow issues in commercial supply, so make sure you are using the right products. Bead-based solutions avoid the use of organic solvents or nucleic acid precipitation, helping to eliminate the problems often encountered with filter-based nucleic acid isolation methods.
For your unique assay or application, we offer a wide range of sample preparation solutions, including Invitrogen Dynabeads magnetic beads, which are customizable for your workflow, or turnkey Applied Biosystems MagMAX kits, which are optimized for easy integration. Both Dynabeads and MagMAX kits work seamlessly with Thermo Scientific KingFisher automation instruments for high-throughput genomic analysis. For more unique or demanding applications, our R&D scientists will work closely with you to customize our beads and buffers to your exact specifications.
Take no chances in the effort to achieve your goals. Partner with us for the quality, security of supply, customization, and expertise you can expect from the world leader in serving science.
Dynabeads products offer many unique benefits and are widely used for specific capture and detection of nucleic acids in clinical samples. Some of the products are from our existing bio-discovery portfolio, while others are developed and supplied on an OEM basis. Our beads are monodisperse, with tight CV (coefficient of variation) specifications and exceptional batch-to-batch reproducibility.
In addition to a broad portfolio of off-the-shelf Dynabeads products, our patented technology and proprietary knowledge enable customized product development.
Customization may include:
Capture | Dynabeads | Surface | Bead size | Characteristics |
Generic | MyOne SILANE | Silica-like | 1.1 µm |
|
Generic | MyOne Carboxylic Acid | Hydrophilic | 1.1 µm |
|
Specific |
| |||
Generic | M-270 Carboxylic Acid | Hydrophilic | 2.8 µm |
|
Specific |
| |||
Specific | MyOne Streptavidin C1 | Hydrophilic | 1.1 µm |
|
Specific | MyOne Streptavidin T1 | Hydrophilic | 1.1 µm |
|
Specific | M-270 Streptavidin | Hydrophilic | 2.8 µm |
|
Specific | M-280 Streptavidin | Hydrophobic | 2.8 µm |
|
Specific | Oligo(dT)25(M-270) | Hydrophilic | 2.8 µm |
|
MagMAX kits offer a turnkey solution for nucleic acid purification for your application, including pathogen detection and formalin-fixed, paraffin-embedded (FFPE) solid tumor biopsies. The MagMAX solution is an integrated system designed to simplify, standardize, and accelerate preparation of samples derived from various sources for downstream molecular analysis.
Nucleic acid isolated | Application | Features | Sample types | |
True total RNA (small RNA– enriched total RNA) | RNA-based oncology or infectious disease testing | Works with most samples of interest, including plasma, serum, whole blood, cell culture, tissue, and urine |
| |
Genomic DNA | Genotyping for inherited disease, oncology, and infectious disease testing | Compatible with common sample types (whole blood, tissue, saliva, buccal swabs, and oral rinse) |
| |
Cell-free DNA | Noninvasive prenatal testing (NIPT), liquid biopsy | Whole blood |
| |
Cell-free DNA | NIPT, liquid biopsy | Whole blood |
| |
DNA and RNA | Genotyping and gene expression for solid tumors | FFPE samples |
| |
RNA and DNA | Infectious disease testing | Serum and plasma; nasal, tracheal, and cloacal swabs; ear samples; whole blood; semen; oral fluid; feces |
| |
Genomic DNA | A variety of molecular protocols, particularly for direct-toconsumer testing | Saliva |
|
KingFisher purification systems are designed to deliver high-quality results with minimal hands-on time, helping you automate a significant part of your workflow. KingFisher systems use permanent magnetic rods and disposable tip combs to collect, transfer and mix magnetic particles.
Watch the automated, medium- to high-throughput Thermo Scientific KingFisher Apex instrument operating to process sample preparation. Building on decades of KingFisher product value and excellence, the KingFisher Apex represents the pinnacle of automated purification for DNA, RNA, proteins, and cells.
KingFisher Duo Prime | KingFisher Flex | KingFisher Apex | KingFisher Presto |
---|---|---|---|
|
|
|
|
Dynabeads products offer many unique benefits and are widely used for specific capture and detection of nucleic acids in clinical samples. Some of the products are from our existing bio-discovery portfolio, while others are developed and supplied on an OEM basis. Our beads are monodisperse, with tight CV (coefficient of variation) specifications and exceptional batch-to-batch reproducibility.
In addition to a broad portfolio of off-the-shelf Dynabeads products, our patented technology and proprietary knowledge enable customized product development.
Customization may include:
Capture | Dynabeads | Surface | Bead size | Characteristics |
Generic | MyOne SILANE | Silica-like | 1.1 µm |
|
Generic | MyOne Carboxylic Acid | Hydrophilic | 1.1 µm |
|
Specific |
| |||
Generic | M-270 Carboxylic Acid | Hydrophilic | 2.8 µm |
|
Specific |
| |||
Specific | MyOne Streptavidin C1 | Hydrophilic | 1.1 µm |
|
Specific | MyOne Streptavidin T1 | Hydrophilic | 1.1 µm |
|
Specific | M-270 Streptavidin | Hydrophilic | 2.8 µm |
|
Specific | M-280 Streptavidin | Hydrophobic | 2.8 µm |
|
Specific | Oligo(dT)25(M-270) | Hydrophilic | 2.8 µm |
|
MagMAX kits offer a turnkey solution for nucleic acid purification for your application, including pathogen detection and formalin-fixed, paraffin-embedded (FFPE) solid tumor biopsies. The MagMAX solution is an integrated system designed to simplify, standardize, and accelerate preparation of samples derived from various sources for downstream molecular analysis.
Nucleic acid isolated | Application | Features | Sample types | |
True total RNA (small RNA– enriched total RNA) | RNA-based oncology or infectious disease testing | Works with most samples of interest, including plasma, serum, whole blood, cell culture, tissue, and urine |
| |
Genomic DNA | Genotyping for inherited disease, oncology, and infectious disease testing | Compatible with common sample types (whole blood, tissue, saliva, buccal swabs, and oral rinse) |
| |
Cell-free DNA | Noninvasive prenatal testing (NIPT), liquid biopsy | Whole blood |
| |
Cell-free DNA | NIPT, liquid biopsy | Whole blood |
| |
DNA and RNA | Genotyping and gene expression for solid tumors | FFPE samples |
| |
RNA and DNA | Infectious disease testing | Serum and plasma; nasal, tracheal, and cloacal swabs; ear samples; whole blood; semen; oral fluid; feces |
| |
Genomic DNA | A variety of molecular protocols, particularly for direct-toconsumer testing | Saliva |
|
KingFisher purification systems are designed to deliver high-quality results with minimal hands-on time, helping you automate a significant part of your workflow. KingFisher systems use permanent magnetic rods and disposable tip combs to collect, transfer and mix magnetic particles.
Watch the automated, medium- to high-throughput Thermo Scientific KingFisher Apex instrument operating to process sample preparation. Building on decades of KingFisher product value and excellence, the KingFisher Apex represents the pinnacle of automated purification for DNA, RNA, proteins, and cells.
KingFisher Duo Prime | KingFisher Flex | KingFisher Apex | KingFisher Presto |
---|---|---|---|