Search Thermo Fisher Scientific
Thermo Fisher Scientific은 통합형 모세관 전기영동 차세대 시퀀싱(CE-NGS) 법의학 workflow 솔루션을 통해 샘플에서 더 많은 해답을 얻도록 지원하기 위해 최선을 다하고 있습니다.
난해한 샘플 또는 일상적인 샘플을 분석하든 간에, 당사의 영업, 서비스, 기술 지원 전문가 3,700명 이상을 적극 활용하면 workflow에 단순성, 확장성, 속도를 더한 답변을 얻을 수 있기 때문에 가장 난해한 사건도 100% 확신하며 분석할 수 있습니다.
Count on our 3,700+ sales, service, and technical support specialists to provide answers that add simplicity, scalability, and speed to your workflow—enabling you to analyze your most difficult cases with complete confidence.
The Applied Biosystems RapidHIT ID System generates lab-quality forensic DNA profiles in only 90 minutes with one minute of hands-on time from single-source samples. Producing STR profiles has never been this fast and easy. The compact, easy-to-use RapidHIT ID System can be set up in a day in virtually any setting, and accompanying Applied Biosystems RapidLINK Software enables real-time access, review and control.
The Applied Biosystems SeqStudio Genetic Analyzer for HID is a 4-capillary, easy-to-use, and convenient benchtop system that delivers gold-standard STR fragment analysis and Sanger sequencing with just one click. It is easily used across a broad range of forensic STR kits and applications. Get the same answers you trust with the outstanding data quality, service, and support you’ve come to expect from Applied Biosystems instruments.
The SeqStudio Flex Genetic Analyzer, available in both an 8- and 24-capillary format, incorporates a built-in touchscreen computer, data collections software, and continuously accessible four-plate deck. The complete system combines the instrument with Applied Biosystems STR kits, reagents, consumables, and GeneMapper ID-X Software v1.7, as well as industry-leading support, providing an integrated HID solution that significantly improves ease of use and application efficiency.
The Applied Biosystems Precision ID NGS System for human identification can help you solve tough cases by getting more information from your challenging samples. By combining the Ion Chef System and either of the Ion GeneStudio S5 next-generation sequencing systems with Applied Biosystems Precision ID panels that include forensically relevant markers, adopting NGS in your laboratory is now simpler than ever.
| Fast and easy-to-use system | Easy-to-use, flexible system | Meets the needs of validated and process-controlled environments | Meets the needs of validated and process-controlled environments with connectivity and flexible scheduling options |
| ||||
| RapidHit ID System | SeqStudio Genetic Analyzer | 3500 Series Genetic Analyzers | SeqStudio Flex Genetic Analyzer |
Number of capillaries/cartridges | 1 | 4 | 8 (3500), 24 (3500xL) | 8 (3500), 24 (3500xL) |
Ease of use | Simple—no pipeting required | Easy, with one click universal cartridge | Moderate—some maintenance required | Moderate—some maintenance required |
Number of dyes | 6 | 8-dye capable | 8-dye capable | 8-dye capable |
RFID | Yes | Yes | Yes | Yes |
Polymer type | Proprietary polymer | POP-1 integrated into click-in cartridge | POP-4, POP-6, POP-7 | POP-4, POP-6, POP-7 |
Sample capacity | 1 | 1 standard 96-well plate or 12 standard 8-strip tubes | 2 sample plates (96-well) | 4 sample plates (96-well) |
Integrated plate stacker | No | No | 2-plate loading deck | 4-plate loading deck |
Applications | HID applications, arrestee testing, crime scene testing, and field testing | HID applications, fragment analysis, and sequencing | HID applications, fragment analysis, and sequencing | HID applications, fragment analysis, and sequencing |
HID applications run time | 90 minutes sample to answer | 39 min | 37 min | 37 min |
Maximum HID applications throughput (samples/day)** | 16 | 147 | 296 (8-cap), 999 (24-cap) | 296 (8-cap), 999 (24-cap) |
HID kits accepted | Sample cartridge includes STR master mix for GlobalFiler Express and NGM Select kits | All 5- and 6-dye STR kits from all vendors | All 5- and 6-dye STR kits from all vendors | All 5- and 6-dye STR kits from all vendors |
Sample type | Casework: RapidINTEL cartridge | Casework: purified DNA | Casework: purified DNA | Casework: purified DNA |
Database: Rapid ACE cartridge | Database: swab, treated or untreated paper | Database: swab, treated or untreated paper | Database: swab, treated or untreated paper | |
Use level of experience | Technician: forensic lab experience not required | Trained forensic scientist | Trained forensic scientist | Trained forensic scientist |
모세관 전기영동용 Applied Biosystems™ 3500 Series는 광학 및 열 하위 시스템 전용 플랫폼 개선 기능 다수를 통합하면서도 업계 최초로 혁신적인 소모품 시스템 접근법을 활용하고 있습니다. 함께 작용하는 이 요소들을 통해 유전자 분석기 성능을 최고 수준으로 극대화할 수 있습니다.
신원감식(HID)용 Applied Biosystems™ Precision ID NGS System은 난해한 샘플에서 더 많은 정보를 확보하여 어려운 사건을 해결하는 데 도움이 될 수 있습니다. Ion Chef™ System과 Ion S5™ 또는 the Ion S5™ XL System을 법의학적 관련 표지자를 포함하는 Applied Biosystems™ Precision ID 패널과 결합함으로써 이제 실험실에 NGS를 그 어느 때보다도 쉽게 도입할 수 있게 되었습니다.
혁신적인 하드웨어 설계, 소모품에 대한 완전히 새로운 접근법, 그리고 강력한 소프트웨어를 통해 3500 Series는 일상적인 작업에 새로운 수준의 성능과 편의성을 더해 줍니다. Applied Biosystems™의 밸리데이션을 거친 신원감식(HID) 시약, 소모품, 소프트웨어뿐 아니라 업계 최고의 지원을 모두 갖춘 3500 Series는 HID 실험실의 생산성과 자신감을 크게 향상시켜 주는 통합 HID 시스템 솔루션입니다.
Applied Biosystems™ GlobalFiler™ STR PCR 증폭 키트는 증폭 시간을 단축하면서도 데이터 회수력을 극대화합니다. 유일하게 완전히 통합된 밸리데이션된 법의학 workflow의 일부인 이 획기적인 6-dye, 24-loci 기술은 전례 없이 우수한 실험실 성능을 통해 적은 수고로 더 많은 성과를 낼 수 있습니다.
Applied Biosystems™ Yfiler™ Plus PCR Amplification Kit도 6-Dye™ 기술을 활용하고 높은 배경의 여성 DNA에 남성 DNA가 미량 존재하기 때문에 증거 샘플을 기존 autosomal STR 분석으로 해결하기 어려운 경우에 사용할 수 있는 중요한 Y-STR 표지자 27개를 포함합니다.
Applied Biosystems™ Precision ID GlobalFiler™ NGS STR Panel은 GlobalFiler PCR Amplification Kit에 있는 Y-indel 및 amelogenin 성별 표지자와 더불어 동일한 autosomal STR 21개를 포함합니다. 이 패널은 SE33을 사용하는 대신 추가 복대립유전자 STR 표지자 9개(총 33개 표적 대상)를 포함하여 복잡한 사건별 작업 샘플의 종합적인 해결에 유용합니다.
The Precision ID GlobalFiler NGS STR Panel is currently available for use with manual library preparation, template preparation on the Ion Chef™ System or Ion OneTouch™ 2 System, and sequencing on the Ion PGM™ System.
Applied Biosystems™ Precision ID mtDNA Whole Genome Panel은 법의학 어플리케이션용으로 특별히 개발된 혁신적인 mtDNA(mitochondrial DNA) 시퀀싱 방법을 사용합니다. 이 mtDNA tiling 접근법에는 각 pool에서 작은 amplicon 81개로 구성된 primer pool 2개가 포함되고, 평균 크기는 163 bp이기 때문에 털줄기, 치아, 골과 같이 크게 손상되고 분해된 샘플에서 최적의 미토콘드리아 게놈 적용 범위 데이터를 확보하는 데 도움이 됩니다.
Precision ID mtDNA Control Region Panel은 Ion AmpliSeq mtDNA Whole Genome Panel에서 사용되는 것과 동일한 mtDNA tiling 접근법을 사용합니다. 이 표적 대조군 영역 패널은 amplicon 7개를 가진 소형 primer pool 세트를 포함하고, 각 세트는 1.2 kb 제어군 영역에 이르므로 HV-I, II, III를 포함합니다.
Applied Biosystems™ Precision ID Ancestry Panel은 생물지리적 가계 정보를 제공하고 조사 절차를 안내할 수 있는 autosomal 표지자(SNP) 165개를 포함합니다.
Applied Biosystems™ Precision ID Identity Panel은 표지자 124개로 구성되어 있고 법의학 분석에서 사용하는 STR 유전형 일치 확률과 유사한 방법으로 개인을 감별하도록 도와줍니다.
Our Human Identification Professional Services (HPS) team has been helping customers like you successfully navigate the validation process required to bring new technologies into operation. At the core of our offerings are services designed to help forensic DNA labs integrate, validate, and implement new methodologies and technologies.
당사의 신원감식 전문 서비스(Human Identification Professional Services, HPS) 팀은 고객이 새 기술을 적용하는 데 필요한 밸리데이션 절차를 성공적으로 진행할 수 있도록 지원하고 있습니다. 당사 서비스의 핵심은 법의학 DNA 실험실이 새로운 방법과 기술을 통합하고, 밸리데이션하고, 실행할 수 있도록 지원하는 서비스입니다.
Crime scene samples can originate from harsh environmental locations, which may result in various stages of degradation and contamination. Imagine a 20-year-old stamp that was stored away in a closet, trace amounts of blood from an exploded IED, a partially eaten piece of pizza, or evidence collected from an unflushed toilet? How would you begin to evaluate these challenging samples?
This webinar details the major updates made at the Washoe County Sheriff's Office Forensic Science Division. DNA Technical Leader Dave Jackson will describe these changes and discuss the improvement in services to the community that is expected to help solve crimes faster.
This webinar presents in-depth evaluations of CE analysis tools designed to minimize hands-on time and time spent on routine data review, enabling more complex sample analysis. In this webinar you’ll learn:
Liz Peters from Fiji Police Force discusses how bringing new forensic DNA technology into her lab has improved sample processing to help solve crime faster.
Forensic scientists at the State Forensic Medicine Service are working to solve one of Lithuania’s greatest historical mysteries—identifying the remains from an 1863 uprising. Marija Caplinskiene, MD, PhD, discusses how her team is using both capillary electrophoresis and next-generation sequencing to analyze DNA from recovered bones found in recently revealed graves.
From understanding how body decomposition affects forensic DNA analysis to simulating IED explosions with the goal of finding touch amounts of DNA, Sheree Hughes-Stamm, PhD from Sam Houston State University describes her work on optimizing DNA extraction and STR analysis methods, and when to apply next-generation sequencing to her studies.
For Research, Forensic, or Paternity Use Only. Not for use in diagnostic procedures.
STR, SNP, mtDNA용 Precision ID 패널은 Ion AmpliSeq™ 기술을 활용합니다. 이 기술은 표적 시퀀싱에 PCR의 간편성을 더할 뿐만 아니라 10~1000개의 게놈 영역을 단일 PCR 튜브에서 동시에 증폭시켜 줍니다. 1ng의 DNA 샘플만으로도 단 5단계의 피펫팅과 45분의 hands-on 시간을 거쳐 법의학 표지자를 빠르고 쉽게 분석할 수 있습니다.
성폭력 사건 샘플 처리 workflow를 능률화하는 방법 알아보기—Alison Holt, Thermo Fisher Scientific
신원감식(HID) 전문 서비스 Workflow 밸리데이션: Quantifiler™ Trio 키트 및 GlobalFiler™ 키트—Joanne B. Sgueglia, Thermo Fisher Scientific
난해한 사건 작업에 대한 Y-STR 데이터 해석 및 법원에 데이터 제출—Lutz Roewer 박사(독일 샤리테 의과대학 법의학 연구소)와 Franz Neuhuber 박사(오스트리아 잘츠부르크 대학교 법의학 연구소)
법의학에 맞게 NGS(next-generation sequencing)를 조정하는 것은 패러다임의 전환이 아니다 —Bruce Budowle 박사, 미국 노스 텍사스 대학교 건강과학센터
NGS 기술을 사용하는 미토콘드리아 DNA 시퀀싱 소개— Walther Parson 박사, 오스트리아 인스브루크 의과대학 법의학 연구소
Ion Torrent™ NGS 자동화 기능을 사용해 법의학적 DNA 분석 가속화—Joseph Chang, Thermo Fisher Scientific
신원감식(HID) 리소스 라이브러리에서 자세히 알아보십시오.