Search Thermo Fisher Scientific
All EVOS microscopes share common features: high resolution imaging; bright LED light sources; high-resolution cameras and optics; compact, efficient design; minimal handling; and powerful, intuitive software. The one that’s right for you depends on your needs and applications. For a versatile laboratory workhorse, choose between the fully automated EVOS M7000 Imaging System and the advanced mechanical EVOS M5000 Imaging System. For a basic microscope suitable for cell culture applications, choose the EVOS M3000 Imaging System.
If your application involves high-content analysis (HCA) or screening, consider the EVOS M7000 HCA Package or the CellInsight platforms.
|
|
|
Model type | ||
Full automation with both monochrome with color cameras for brightfield, phase contrast, color, and fluorescent imaging | Semi-automation with both monochrome with color image capture for brightfield, phase contrast, color, and fluorescent imaging | Fixed stage for brightfield, phase contrast, color, and fluorescent imaging |
Description | ||
The EVOS M7000 has been designed with advanced capabilities to simplify demanding slide and cell-based imaging applications including live-cell analysis, image tiling, and Z-stacking. | The EVOS M5000 is designed to support a broad range of applications, from quick check to acquisition of publication-quality images. The Stage View feature allows you to effortlessly locate, track, and record points of interest. | The EVOS M3000 is ideal for routine cell culture applications including confluency, fluorescent protein expression, sample QC and stem cell monitoring. |
EVOS M7000 | EVOS M5000 | EVOS M3000 |
---|---|---|
Simple Installation | ||
Installation and training | ||
Service Team | User | User |
Motorized encoded X/Y scanning stage | ||
--- | --- | |
Manual mechanical stage (optional) | ||
--- | ||
Objective turret positions | ||
5 | 5 | 4 |
Phase contrast | ||
Objective range | ||
1.25-100x | 1.25-100x | 1.25- 60X |
Fluorescence channels | ||
2 | ||
Fluorescence LED light cubes | ||
Monochrome or color camera option (built-in, not optional) | ||
Both | Mono with LED-based RGB illumination scheme | Color camera |
Epifluorescence images | ||
Transmitted-light images | ||
Color images | ||
Benchtop system | ||
Suitable for use in tissue culture hood | ||
--- | ||
Darkroom needed | ||
No | No | No |
Onstage incubator for time-lapse imaging | ||
Optional | Optional | --- |
Autofocus | ||
Multichannel | Multichannel | --- |
Z-stack capability | ||
--- | ||
Automated multiwell plate screening | ||
--- | --- | |
Cloud connectivity | ||
USB Ports | ||
External monitor support | ||