microscopic image of bacteria
Optimized for use in bacterial cells, our BacLight series of bacterial viability and vitality assays for flow cytometry allow clear separation of living and dead cells based on various cellular characteristics. Due to the inherent differences between prokaryotic and eukaryotic cells, specialized dyes must be used to differentiate live and dead cell populations in bacteria.
  • Ease of use—Staining in 5–10 minutes with no wash steps.
  • Specific—Different colors for live and dead cells allows easy separation even in mixed populations.
  • Reliable—Consistent results regardless of bacterial species
  • Versitile—Also compatible with other detection platforms, including fluorescent microscopes, fluorometers and microplate readers
Membrane Integrity Viability Kit
The LIVE/DEAD BacLight Bacterial Viability Kit for flow cytometry is convenient and easy-to-use for monitoring the viability of bacterial populations as a function of the membrane integrity of the cell. Cells with a compromised membrane that are considered to be dead or dying will stain red, whereas cells with an intact membrane will stain green.
Figure 1. Micrococcus luteus and Bacillus cereus stained with the LIVE/DEAD BacLight Bacterial Viability Kit for flow cytometry. When incubated with the SYTO 9 and propidium iodide nucleic acid stains provided in this kit, live bacteria with intact cell membranes fluoresce green and dead bacteria with compromised membranes fluoresce red.
ProductLaserEx/Em*Live cell stainDead cell stainCat. No.
LIVE/DEAD BacLight Bacterial Viability Kit488 nm480/500 nm (L),
490/635 nm (D)
SYTO9Propidium IodideL7012
*Indicates excitation and emission. L = live, D = dead.
Metabolic activity viability kits
The BacLight RedoxSensor Bacterial Vitality Assay for flow cytometry provides effective reagents for evaluating bacterial cell health and vitality based on detecting activity of bacterial oxidases and reductases. Enhanced fluorescent signal is observed in bacterial cells with active metabolic processes. The BacLight RedoxSensor Bacterial Vitality kits also withstand fixation procedures.
Staphylococcus aureus cells were stained with RedoxSensor Green reagent and propidium iodide
Figure 2.
BacLight RedoxSensor Green Bacterial Vitality Kit. A mixture of healthy and killed Staphylococcus aureus cells were stained with 100 nM RedoxSensor Green reagent and propidium iodide. Dual-color flow cytometric analysis of the sample, gated on bacteria using scatter, shows both the healthy and the membrane-compromised cell populations. Similar results can be obtained with Escherichia coli.
Citations
See citations for the BacLight RedoxSensor Green Bacteria Vitality Kit
See citations for the BacLight RedoxSensor CTC Vitality Kit
ProductLaserEx/Em*Live cell stainDead cell stainCat. No.
BacLight RedoxSensor Green Bacteria Vitality Kit488 nm490/520 nm (L),
490/635 nm (D)
SYTO9Propidium IodideB34954
BacLight RedoxSensor CTC Vitality Kit488 nm480/500 nm (L),
490/635 nm (D)
SYTO24Propidium IodideB34956
*Indicates excitation and emission. L = live, D = dead.
Bacterial Membrane Potential Kit
The BacLight Bacterial Membrane Potential Kit for flow cytometry provides the fluorescent membrane-potential indicator dye, DiOC2(3), which exhibits green fluorescence in all bacterial cells, but it becomes more concentrated in healthy cells that are maintaining a membrane potential, causing the dye to self-associate and the fluorescence emission to shift to red. The red- and green-fluorescent bacterial populations are easily distinguished using a flow cytometer.
Fluorescence dot plot showing Staphylococcus aureus incubated with DiOC2(3)
Figure 3. BacLight Bacterial Membrane Potential Kit for flow cytometry. The red vs. green fluorescence dot plot show Staphylococcus aureus incubated with 30 µM DiOC2(3) for 30 minutes in either the presence or absence of 5 µM carbonyl cyanide 3-chlorophenylhydrazone (CCCP). Flow cytometer data were collected with log amplification.
Citations
See citations for the BacLight Bacterial Membrane Potential Kit
ProductLaserEx/Em*Live cell stainDead cell stainCat. No.
BacLight Bacterial Membrane Potential Kit488 nm482/497 nmDiOC2(3)NoneB34950
*Indicates excitation and emission.
Resources
Fluorophore and reagent selection guide for flow cytometry
Download Flow Cytometry Protocols Handbook
Spectral Flow Cytometry Fundamentals
Invitrogen eBioscience Resources—Selection guides, Best Protocols, product performance and more.
Intracellular Staining for Flow Cytometry How-To Video—for detecting cytokines and intranuclear markers.
Flow Cytometry Learning Center—Access flow cytometry educational resources for better experiment planning and execution.
Flow Cytometry Panel Builder—Design your flow cytometry panel with this online tool for a simplified, customizable experience to fit your needs.
5 Steps Resources
Support
Flow Cytometry Support Center—Find technical support recommendations for your flow cytometry workflows, including tips for experimental setup and in-depth troubleshooting help.
Flow Cytometry Panel Design Support—Work with one of our technical sales specialists to discuss your experimental needs and guide you through the process.
Flow Cytometry Protocols
Flow Cytometry Application Notes and Posters
Contact us