Search Thermo Fisher Scientific
MagMAX Viral/Pathogen kits and reagents use magnetic bead-based technology to purify high-quality nucleic acid from a range of research sample types both manually and through automation. To help save time and increase reproducibility, combine with KingFisher instruments for automated purification. Our kits are recommended for viral nucleic acid isolation for SARS-CoV-2 and other other infectious disease pathogens.
The following products are recommended for research based viral nucleic acid isolation for SARS-CoV-2 and other infectious disease pathogens. To learn about the differences between the kits, please see Frequently Asked Questions below.
For more information on ordering MagMAX Viral/Pathogen II Nucleic Acid Isolation kit (A48383R) please contact customer service.
Designed for efficient extraction of DNA and RNA from a diverse range of human biological specimens (including hard to lyse sample types) for infectious disease research applications.
MagMAX Viral/Pathogen Nucleic Acid Isolation Kit (MVP I) Order now |
| MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit Order now | |
Who is it for? | These MagMAX Viral/Pathogen Nucleic Acid Isolation Kits are designed for use by labs that are that are researching SARS-CoV-2 and other infectious disease pathogens. | ||
Sample types | MVP I and MVP II: Universal viral transport media (VTM), bronchoalveolar lavage (BAL) and other respiratory research samples, serum, plasma, urine, CSF, whole blood, and saliva | MVP Ultra: Blood, Urine, Swabs, Lavage, Universal viral transport media (VTM), ThinPrep and Eswab | |
Automation level | Automation-ready protocols designed for Thermo Scientific KingFisher Apex, KingFisher Flex and KingFisher Duo Prime systems | ||
Sample input volume | 200 µL of VTM sample recommended | Sample volume inputs from 200 µL to 2 mL of recommended sample | |
Elution volume | 50 μL | Elution volumes ranging from 50 to 100 µL | |
Time per run | MVP I and MVP II: 96 samples can be processed in <40 minutes | MVP Ultra: 96 samples can be processed in <60 minutes | |
Protocol update |
| — | |
What is the difference between the two kits? | Binding Beads: In order to support the unprecedented demand for RNA extraction products for COVID-19 testing, there has been a raw material change in the bead component contained in the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit. The MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit contains these beads. The chemistry driving the RNA to the beads is identical, and the nucleic acid binding capacity of each bead type is sufficient to provide similar results between the two products. Bulk reagent format: The MagMAX Viral/Pathogen II kit is only available in bulk prep format with 1,000 or 2,000 reactions. | The MVP Ultra kit includes a proprietary enzyme mix that enables lysis and nucleic acid recovery from the toughest pathogens, such as gram-positive bacteria, yeast, and fungi. | |
Cat. No. | A48310 (up to 2,000 preps/kit) This kit contains the following SKUs: A42359, A42360, A42362, A42363, A42364 | A48383R (up to 2,000 preps/kit) | A42356 (up to 100 preps) |
A42352 (up to 200 preps/kit) |
The following protocols can be used for either the MagMAX Viral/Pathogen Kit (MVP I), MagMAX Viral/Pathogen II Kit (MVP II) and MagMAX Viral/Pathogen Ultra Kit for extraction of SARS-CoV-2 and other infectious disease pathogens, which supports sample input volume from 5 mL to 200 uL and reduction from 3 wash steps to 2 wash steps. Protocols can be downloaded for the KingFisher Flex 96 Deep-well head (5400630).
Protocol type | Standard wash (3 wash steps) | Reduced wash (2 wash steps) |
---|---|---|
200 μL script | MVP_Flex_200ul.bdz | MVP_2Wash_200_Flex.bdz |
400 μL script | MVP_Flex_400ul.bdz | MVP_2Wash_400_Flex.bdz |
200–400 µL Script | MVP Ultra_200ul_400ul | N/A |
500 µL–2 mL | MVP Ultra_500ul_2ml | N/A |
Extract and analyze viral nucleic acid directly from a saliva sample. The following protocol can be used for either MagMAX Viral/Pathogen (MVP I) and MagMAX Viral/Pathogen II (MVP II) for extraction of SARS-CoV-2 and other infectious disease pathogens, which support the isolation of nucleic acid from 200 uL of saliva (raw and preserved).
Want to isolate without an instrument? See our manual protocol/user guide for use with magnetic stands.
Figure 1. Viral Extraction Kit Comparison - RNA Virus. Nucleic acid was extracted using the MagMAX Viral/Pathogen Nucleic Isolation Kit and competitor kits from 5 different matrices spiked with inactivated viral particles Once extracted, one-step qRT-PCR was performed on 1ul of each sample in duplicate to determine performance between the kits. Panel A: (-) ssRNA Virus. Panel B: (+) ssRNA Virus
Figure 2: Viral Extraction Kit Comparison - DNA Virus. Nucleic acid was extracted using the MagMAX Viral/Pathogen Nucleic Isolation Kit and and competitor kits from 5 different matrices spiked with inactivated viral particles. Once extracted, one-step qRT-PCR was performed on 1ul of each sample in duplicate to determine performance between the kits.
A. The MagMAX Prime Viral/Pathogen NA Isolation Kit offers several key benefits. Firstly, it utilizes our most advanced formulation, guaranteeing maximum nucleic acid yield and enhancing assay sensitivity. Additionally, it enables users to conveniently process various sample types on a single plate using the same KingFisher script. Lastly, this kit is compatible with the KingFisher Flex, Apex, Apex Dx (RUO mode), and Presto sample purification instruments, providing an automated workflow for seamless integration.
A. There are two key differences between the MVP I and MVP II products:
Binding Beads: A proprietary raw material change in the bead component is different. The chemistry driving the RNA to the beads is identical and the nucleic acid binding capacity provides similar/equivalent results between the two products. Bulk reagent format: The second difference is that the MVP II (A48383R) product is only available in the bulk format. There is no 100-reaction option for the MVP II kit.
A. Yes, we have a manual ready protocol and user guide for use with a magnetic stand.
A. Yes, the product contents (Binding Solution, Wash Buffer, Proteinase K, and Elution Solution formulations) are the same in both kits.
Yes, the overall workflow, in terms of number of steps and volume usage, remains the same; if you have been using the MVP I (A42353, A48310) kit you should not have to change your protocol.
IMPORTANT: The protocol for both the MVP I and MVP II products has been updated to include a separate lower input option: You can now use a sample input volume of 200 μL (vs. the standard 400 μL) with a reduced (halved) amount of extraction reagents. What does this mean? It means that you can now get 2,000 preps from your MVP I (A48310) or MVP II (A48383R) products. This also means that the smaller pack size of the MVP I kit (A42352) can support 200 preps.
A. Yes, there will be a new script for the 200 μL sample input protocol and both kit versions can be used with it.
A. Yes, all the same samples are compatible.
A. Yes. You can use the MVP II (A48383R) product just as you would use the MVP I (A42352, A48310) product with the TaqPath COVID-19 Combo Kit. When using the 200 μL sample input protocol, please note that the eluate input into the PCR will be changed to 10 uL (5uL eluate input when using 400 μL).
A. No, the MVP II is NOT replacing the existing MagMAX Viral/Pathogen Kit (Cat. Nos. A423352, A48310). The intent of this product to expand the number of samples that can be processed with our MagMAX reagents.
A. Yes, they are both manufactured under the same ISO 13485 standards which utilizes a high set of quality control measures.
A: No, the bulk reagents for both kits are the same price.
A: For the 96DW plate format: 200-400 μL sample volume input (200 μL for whole blood).
For the 24DW plate format: 500-2000 μL sample volume input (1000 μL for whole blood).
A: Human biofluid samples (e.g. blood, serum, plasma, urine, CSF, lavage, saliva), samples stored in transport media (e.g. BD® Universal Viral Transport media), and bacteriophage culture.
A: Nucleic acid has been successfully detected from as low as 50 viral copies in the starting sample.
A: (-)ssRNA virus, (+)ssRNA Virus, ssDNA virus, dsDNA virus, Gram negative bacteria.
A: qRT-PCR, qPCR, sequencing.
A: 30 min from sample to extracted nucleic acid for up to 96 samples using the MVP I and MVP II kits. 65 min from sample to extracted nucleic acid for up to 96 samples using the MVP Ultra kit.
A: Kit is compatible with all KingFisher instruments (Flex, Duo Prime, and Presto) and can also be used for manual processing in 96-well plate.
A: MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit (A42356) is recommended for all variations of the TrueMark panels, especially if a single extraction kit is preferred for a simplified workflow. This kit is especially effective for difficult to lyse samples containing gram-positive bacteria, yeast, or fungi pathogens.
Alternative workflows using MagMAX Viral/Pathogen I (MVP I) Nucleic Acid Isolation Kit (A42352, A48310) and MagMAX Viral/Pathogen (MVP II) Nucleic Acid Isolation Kit (A48383R) can also be used with the TrueMark panels.
A: MagMAX Viral/Pathogen (MVP II) Nucleic Acid Isolation Kit (A48383R) is recommended for both feasibility and effectiveness to be utilized with the TrueMark STI Select Panels in detection of high-incidence sexually transmitted pathogens.
Alternatively, MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit (A42356) is also compatible with the TrueMark STI Select Panels.
To help save time and increase reproducibility with the MagMAX Viral/Pathogen Nucleic Acid Isolation kits, combine with E1-ClipTip Equalizer electronic pipettes and Multidrop Combi instruments.
Adjustable spacing, Bluetooth enabled electronic pipette. Learn more
Automated reagent dispenser. Learn more
Watch these how-to videos to learn how to use the MagMAX Viral/Pathogen Nucleic Acid Isolation kits with KingFisher Instruments and saliva collection devices.
Watch how to extract RNA for for SARS CoV-2 sample preparation using the MagMAX Viral/Pathogen Nucleic Acid Isolation kit with the KingFisher Flex Purification System for high-throughput nucleic acid isolation.
Watch how to extract RNA for SARS CoV-2 sample preparation using the MagMAX Viral/Pathogen Nucleic Acid Isolation kit with the KingFisher Duo Prime Purification System for medium-throughput nucleic acid isolation.
Watch how to extract RNA for SARS-CoV-2 sample preparation using MagMAX Viral/Pathogen Isolation Kit using the manual extraction process.
Watch this video to learn techniques on how to collect saliva effectively for nucleic acid isolation using the MagMAX Viral/Pathogen Nucleic Acid Isolation kit.
Watch this video to learn techniques on how to collect saliva effectively for nucleic acid isolation using the MagMAX Viral/Pathogen Nucleic Acid Isolation kit.
Watch this video to learn techniques on how to collect saliva effectively for nucleic acid isolation using the MagMAX Viral/Pathogen Nucleic Acid Isolation kit.
Watch this video to learn techniques on how to collect saliva effectively for nucleic acid isolation using the MagMAX Viral/Pathogen Nucleic Acid Isolation kit.
MagMAX Viral/Pathogen Nucleic Acid Isolation Kit (MVP I) Order now |
| MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit Order now | |
Who is it for? | These MagMAX Viral/Pathogen Nucleic Acid Isolation Kits are designed for use by labs that are that are researching SARS-CoV-2 and other infectious disease pathogens. | ||
Sample types | MVP I and MVP II: Universal viral transport media (VTM), bronchoalveolar lavage (BAL) and other respiratory research samples, serum, plasma, urine, CSF, whole blood, and saliva | MVP Ultra: Blood, Urine, Swabs, Lavage, Universal viral transport media (VTM), ThinPrep and Eswab | |
Automation level | Automation-ready protocols designed for Thermo Scientific KingFisher Apex, KingFisher Flex and KingFisher Duo Prime systems | ||
Sample input volume | 200 µL of VTM sample recommended | Sample volume inputs from 200 µL to 2 mL of recommended sample | |
Elution volume | 50 μL | Elution volumes ranging from 50 to 100 µL | |
Time per run | MVP I and MVP II: 96 samples can be processed in <40 minutes | MVP Ultra: 96 samples can be processed in <60 minutes | |
Protocol update |
| — | |
What is the difference between the two kits? | Binding Beads: In order to support the unprecedented demand for RNA extraction products for COVID-19 testing, there has been a raw material change in the bead component contained in the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit. The MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit contains these beads. The chemistry driving the RNA to the beads is identical, and the nucleic acid binding capacity of each bead type is sufficient to provide similar results between the two products. Bulk reagent format: The MagMAX Viral/Pathogen II kit is only available in bulk prep format with 1,000 or 2,000 reactions. | The MVP Ultra kit includes a proprietary enzyme mix that enables lysis and nucleic acid recovery from the toughest pathogens, such as gram-positive bacteria, yeast, and fungi. | |
Cat. No. | A48310 (up to 2,000 preps/kit) This kit contains the following SKUs: A42359, A42360, A42362, A42363, A42364 | A48383R (up to 2,000 preps/kit) | A42356 (up to 100 preps) |
A42352 (up to 200 preps/kit) |
The following protocols can be used for either the MagMAX Viral/Pathogen Kit (MVP I), MagMAX Viral/Pathogen II Kit (MVP II) and MagMAX Viral/Pathogen Ultra Kit for extraction of SARS-CoV-2 and other infectious disease pathogens, which supports sample input volume from 5 mL to 200 uL and reduction from 3 wash steps to 2 wash steps. Protocols can be downloaded for the KingFisher Flex 96 Deep-well head (5400630).
Protocol type | Standard wash (3 wash steps) | Reduced wash (2 wash steps) |
---|---|---|
200 μL script | MVP_Flex_200ul.bdz | MVP_2Wash_200_Flex.bdz |
400 μL script | MVP_Flex_400ul.bdz | MVP_2Wash_400_Flex.bdz |
200–400 µL Script | MVP Ultra_200ul_400ul | N/A |
500 µL–2 mL | MVP Ultra_500ul_2ml | N/A |
Extract and analyze viral nucleic acid directly from a saliva sample. The following protocol can be used for either MagMAX Viral/Pathogen (MVP I) and MagMAX Viral/Pathogen II (MVP II) for extraction of SARS-CoV-2 and other infectious disease pathogens, which support the isolation of nucleic acid from 200 uL of saliva (raw and preserved).
Want to isolate without an instrument? See our manual protocol/user guide for use with magnetic stands.
Figure 1. Viral Extraction Kit Comparison - RNA Virus. Nucleic acid was extracted using the MagMAX Viral/Pathogen Nucleic Isolation Kit and competitor kits from 5 different matrices spiked with inactivated viral particles Once extracted, one-step qRT-PCR was performed on 1ul of each sample in duplicate to determine performance between the kits. Panel A: (-) ssRNA Virus. Panel B: (+) ssRNA Virus
Figure 2: Viral Extraction Kit Comparison - DNA Virus. Nucleic acid was extracted using the MagMAX Viral/Pathogen Nucleic Isolation Kit and and competitor kits from 5 different matrices spiked with inactivated viral particles. Once extracted, one-step qRT-PCR was performed on 1ul of each sample in duplicate to determine performance between the kits.
A. The MagMAX Prime Viral/Pathogen NA Isolation Kit offers several key benefits. Firstly, it utilizes our most advanced formulation, guaranteeing maximum nucleic acid yield and enhancing assay sensitivity. Additionally, it enables users to conveniently process various sample types on a single plate using the same KingFisher script. Lastly, this kit is compatible with the KingFisher Flex, Apex, Apex Dx (RUO mode), and Presto sample purification instruments, providing an automated workflow for seamless integration.
A. There are two key differences between the MVP I and MVP II products:
Binding Beads: A proprietary raw material change in the bead component is different. The chemistry driving the RNA to the beads is identical and the nucleic acid binding capacity provides similar/equivalent results between the two products. Bulk reagent format: The second difference is that the MVP II (A48383R) product is only available in the bulk format. There is no 100-reaction option for the MVP II kit.
A. Yes, we have a manual ready protocol and user guide for use with a magnetic stand.
A. Yes, the product contents (Binding Solution, Wash Buffer, Proteinase K, and Elution Solution formulations) are the same in both kits.
Yes, the overall workflow, in terms of number of steps and volume usage, remains the same; if you have been using the MVP I (A42353, A48310) kit you should not have to change your protocol.
IMPORTANT: The protocol for both the MVP I and MVP II products has been updated to include a separate lower input option: You can now use a sample input volume of 200 μL (vs. the standard 400 μL) with a reduced (halved) amount of extraction reagents. What does this mean? It means that you can now get 2,000 preps from your MVP I (A48310) or MVP II (A48383R) products. This also means that the smaller pack size of the MVP I kit (A42352) can support 200 preps.
A. Yes, there will be a new script for the 200 μL sample input protocol and both kit versions can be used with it.
A. Yes, all the same samples are compatible.
A. Yes. You can use the MVP II (A48383R) product just as you would use the MVP I (A42352, A48310) product with the TaqPath COVID-19 Combo Kit. When using the 200 μL sample input protocol, please note that the eluate input into the PCR will be changed to 10 uL (5uL eluate input when using 400 μL).
A. No, the MVP II is NOT replacing the existing MagMAX Viral/Pathogen Kit (Cat. Nos. A423352, A48310). The intent of this product to expand the number of samples that can be processed with our MagMAX reagents.
A. Yes, they are both manufactured under the same ISO 13485 standards which utilizes a high set of quality control measures.
A: No, the bulk reagents for both kits are the same price.
A: For the 96DW plate format: 200-400 μL sample volume input (200 μL for whole blood).
For the 24DW plate format: 500-2000 μL sample volume input (1000 μL for whole blood).
A: Human biofluid samples (e.g. blood, serum, plasma, urine, CSF, lavage, saliva), samples stored in transport media (e.g. BD® Universal Viral Transport media), and bacteriophage culture.
A: Nucleic acid has been successfully detected from as low as 50 viral copies in the starting sample.
A: (-)ssRNA virus, (+)ssRNA Virus, ssDNA virus, dsDNA virus, Gram negative bacteria.
A: qRT-PCR, qPCR, sequencing.
A: 30 min from sample to extracted nucleic acid for up to 96 samples using the MVP I and MVP II kits. 65 min from sample to extracted nucleic acid for up to 96 samples using the MVP Ultra kit.
A: Kit is compatible with all KingFisher instruments (Flex, Duo Prime, and Presto) and can also be used for manual processing in 96-well plate.
A: MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit (A42356) is recommended for all variations of the TrueMark panels, especially if a single extraction kit is preferred for a simplified workflow. This kit is especially effective for difficult to lyse samples containing gram-positive bacteria, yeast, or fungi pathogens.
Alternative workflows using MagMAX Viral/Pathogen I (MVP I) Nucleic Acid Isolation Kit (A42352, A48310) and MagMAX Viral/Pathogen (MVP II) Nucleic Acid Isolation Kit (A48383R) can also be used with the TrueMark panels.
A: MagMAX Viral/Pathogen (MVP II) Nucleic Acid Isolation Kit (A48383R) is recommended for both feasibility and effectiveness to be utilized with the TrueMark STI Select Panels in detection of high-incidence sexually transmitted pathogens.
Alternatively, MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit (A42356) is also compatible with the TrueMark STI Select Panels.
To help save time and increase reproducibility with the MagMAX Viral/Pathogen Nucleic Acid Isolation kits, combine with E1-ClipTip Equalizer electronic pipettes and Multidrop Combi instruments.
Adjustable spacing, Bluetooth enabled electronic pipette. Learn more
Automated reagent dispenser. Learn more
Watch these how-to videos to learn how to use the MagMAX Viral/Pathogen Nucleic Acid Isolation kits with KingFisher Instruments and saliva collection devices.
Watch how to extract RNA for for SARS CoV-2 sample preparation using the MagMAX Viral/Pathogen Nucleic Acid Isolation kit with the KingFisher Flex Purification System for high-throughput nucleic acid isolation.
Watch how to extract RNA for SARS CoV-2 sample preparation using the MagMAX Viral/Pathogen Nucleic Acid Isolation kit with the KingFisher Duo Prime Purification System for medium-throughput nucleic acid isolation.
Watch how to extract RNA for SARS-CoV-2 sample preparation using MagMAX Viral/Pathogen Isolation Kit using the manual extraction process.
Watch this video to learn techniques on how to collect saliva effectively for nucleic acid isolation using the MagMAX Viral/Pathogen Nucleic Acid Isolation kit.
Watch this video to learn techniques on how to collect saliva effectively for nucleic acid isolation using the MagMAX Viral/Pathogen Nucleic Acid Isolation kit.
Watch this video to learn techniques on how to collect saliva effectively for nucleic acid isolation using the MagMAX Viral/Pathogen Nucleic Acid Isolation kit.
Watch this video to learn techniques on how to collect saliva effectively for nucleic acid isolation using the MagMAX Viral/Pathogen Nucleic Acid Isolation kit.
Find everyday lab essentials to power your PCR every step of the way. Shop now
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications.
© 2023 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.