Search Thermo Fisher Scientific
Isolation (1-3) or depletion (4,5) of murine B cells directly from spleen or lymph node cell suspensions or other samples containing B cells. For rapid and consistent results in protein or gene expression analysis, lyse the B cells while still attached to the beads and directly process for further molecular analysis.
Principle of Isolation
Dynabeads are mixed with the cell sample in a tube. The Dynabeads will bind to the target cells during a short incubation, and then the beadbound cells are separated by a magnet.
Description of Materials
Dynabeads Mouse pan B (B220) are uniform, superparamagnetic polystyrene beads (4.5 μm diameter) coated with a monoclonal antibody specific for the B220 antigen. B220 (also known as CD45R) is expressed on all B cells throughout development, from the early pro-B stage of B cell differentiation (6).
Materials Supplied
5 ml Dynabeads Mouse pan B (B220)
4 x 108 Dynabeads/ml in phosphate buffered saline (PBS), pH 7.4, with 0.1% bovine serum albumin (BSA) and 0.02% sodium azide (NaN3).
The product will process up to 1 x 109 leucocytes (f.ex. splenocytes).
Additional Materials Required
Dynabeads Washing Procedure
Dynabeads should be washed before use.
1. Resuspend the Dynabeads in the vial.
2. Transfer the desired volume of Dynabeads to a tube.
3. Add the same volume of PBS w/0.1 % BSA (or at least 1 ml) and mix.
4. Place the tube in a magnet for 1 min and discard the supernatant.
5. Remove the tube from the magnet and resuspend the Dynabeads to the original volume (step 2) using buffer.
2.2 Sample Preparation
Mouse cells are generally obtained from spleen, thymus or lymph node, but other sources can also be used.
Please visit www.lifetechnologies.com/cellisolation and follow our QuickLinks for recommended sample preparation procedures.
Important Notes
Table 1: Volume of Dynabeads added per 1 ml sample. The volumes can be scaled up as required.
Positive isolation | Depletion | |
---|---|---|
Volume of cells (up to 5 x 106 leucocytes/ml) | 1 ml | 1 ml |
Volume of Dynabeads per 5 x 106 leucocytes | 25 μl | 50 μl |
Total no. of leucocytes processed per product | 1 x 109 | 5 x 108 |
2.3 Depletion or Positive Isolation of B positive Mouse Cells
Invitrogen Dynal AS complies with the Quality System Standards ISO 9001:2000 and ISO 13485:2003.
Description of Materials
Dynabeads FlowComp™ are uniform, superparamagnetic beads (2.8 μm in diameter). Supplied at a concentration of approx. 1 × 109 beads (10 mg) per ml in phosphate buffered saline (PBS), pH 7.4, containing 0.1% bovine serum albumin (BSA) and 0.02% sodium azide (NaN3) as preservatives.
Storage/Stability
This product is stable until the expiry date stated on the label when stored unopened at 2-8°C. Store opened vials at 2-8°C and avoid bacterial contamination. Keep Dynabeads in liquid suspension during storage and all handling steps, as drying will result in reduced performance. Resuspend well before use.
Warnings And Limitations
This product is for research use only. Not intended for any animal or human therapeutic or diagnostic use unless otherwise stated. Follow appropriate laboratory guidelines. This product contains 0.02% sodium azide as a preservative, which is cytotoxic.
Avoid pipetting by mouth!
Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. When disposing through plumbing drains, flush with large volumes of water to prevent azide build up. Certificate of Analysis (CoA) is available upon request. Material Safety Data Sheet (MSDS) is available at .