Search Thermo Fisher Scientific
Epitope tag antibodies are very commonly used to study localization of a target protein in immunofluorescence (IF) assays. An epitope tag can either be fused at C- or N-terminus of a target gene and expressed in a desired expression system.
|
|
Remember to have untransfected, vector alone, and lipofectamine alone samples as controls when testing a construct for the first time.
|
|
|
|
|
|
|
|
|
Equal concentrations of epitope tag antibody should be added to untransfected/transfected samples and any other controls.
For co-localization studies:
For primary conjugates:
No secondary antibody is needed. If you are using a fluorophore-conjugated epitope tag antibody, it is extremely important to have an isotype control. For IF, an isotype control would be a host-matched isotype conjugated with the same fluorophore as the tag antibody. This will help you assess the background signal.
|
|
|
Capture images of untransfected, transfected, no primary/isotype controls at the same exposure settings to compare signal/noise ratios.
For Research Use Only. Not for use in diagnostic procedures.