Search Thermo Fisher Scientific
Epitope tag antibodies are commonly used to study the interactions of the protein of interest with other proteins or nucleic acids. In the case of immunoprecipitation (IP), the protein of interest is expressed as a fusion protein with a tag cloned into either the N- or C-terminus. A tag-specific antibody that is immobilized onto a solid support is used to pulldown the protein of interest. Further confirmation is done by western blotting and/or other assay techniques.
Chromatin immunoprecipitation (ChIP) assays are performed to identify regions of the genome with which DNA-binding proteins, such as transcription factors and histones, associate. In ChIP assays, proteins bound to DNA are temporarily crosslinked and the DNA is sheared prior to cell lysis. The target proteins are immunoprecipitated along with the crosslinked nucleotide sequences which is then identified/analyzed by PCR, sequencing, microarrays, etc.
|
|
|
|
|
Preparation of the Dynabeads-Tag Antibody complex involves incubation of Dynabeads-Protein A/G with the epitope tag antibody. The amount of antibody, Dynabeads, and incubation time required needs to be determined on a case-by-case basis. A good starting point is a 10-minute incubation with 1-10 µg of tag antibody and 50 µL of Dynabeads-Protein A/G.
Specificity of Protein A or G to different Ig classes and subtypes varies and should be carefully considered before selecting an appropriate kit.
IPs should be done with a matched-isotype control antibody to rule out specificity-related issues.
|
|
|
|
|
|
For immunoprecipitation of the target antigen, the Dynabeads-Protein A/G-Tag Antibody complex is incubated with the lysate that contains the target fusion protein leading to formation of the Dynabeads-Protein A/G-Tag Antibody-Fusion Protein immune complex. While the amount of lysate used for this purpose and the incubation time needs to be calibrated on a case-by-case basis, the Dynabeads Protein A or G IP kit recommends 100-1,000 µL of lysate and a 10-minute incubation time as a starting point for optimization.
A sufficient volume of untransfected and transfected lysates (~5-10% of input) should be kept aside to be used as controls in downstream analysis.
|
|
|
|
|
|
|
The analysis of immunoprecipitation is usually done by western blot (use a protocol that is familiar or learn more in the Protein Gel Electrophoresis and Western Blotting Education Center).
However, one of the chief concerns regarding immunoprecipitation detection by western blot is the recognition of prominent antibody light (~25 kDa) and heavy chain (~55 kDa) bands. To circumvent this, it is recommended to use Clean-Blot IP Detection Reagent (HRP) instead for standard HRP conjugated secondary antibodies. This reagent is optimized for post-immunoprecipitation western blot detection of primary antibodies without interference from denatured IP antibody fragments.
|
|
|
|
|
|
|
|
|
|
Make sure that the caps are tightly locked to prevent evaporation of water.
|
|
|
|
|
|
|
|
|
|
|
|
|
Reaction Mixture for 1 sample is as follows:
|
|
|
|
|
|
|
|
|
|
|
|
Preparation of the Dynabeads-Tag Antibody complex involves incubation of Dynabeads-Protein A/G with the epitope tag antibody. The amount of antibody, Dynabeads, and incubation time required needs to be determined on a case-by-case basis. A good starting point is a 10-minute incubation with 1-10 µg of tag antibody and 50 µL of Dynabeads-Protein A/G.
Specificity of Protein A or G to different Ig classes and subtypes varies and should be carefully considered before selecting an appropriate kit.
IPs should be done with a matched-isotype control antibody to rule out specificity-related issues.
|
|
|
|
|
|
For immunoprecipitation of the target antigen, the Dynabeads-Protein A/G-Tag Antibody complex is incubated with the lysate that contains the target fusion protein leading to formation of the Dynabeads-Protein A/G-Tag Antibody-Fusion Protein immune complex. While the amount of lysate used for this purpose and the incubation time needs to be calibrated on a case-by-case basis, the Dynabeads Protein A or G IP kit recommends 100-1,000 µL of lysate and a 10-minute incubation time as a starting point for optimization.
A sufficient volume of untransfected and transfected lysates (~5-10% of input) should be kept aside to be used as controls in downstream analysis.
|
|
|
|
|
|
|
The analysis of immunoprecipitation is usually done by western blot (use a protocol that is familiar or learn more in the Protein Gel Electrophoresis and Western Blotting Education Center).
However, one of the chief concerns regarding immunoprecipitation detection by western blot is the recognition of prominent antibody light (~25 kDa) and heavy chain (~55 kDa) bands. To circumvent this, it is recommended to use Clean-Blot IP Detection Reagent (HRP) instead for standard HRP conjugated secondary antibodies. This reagent is optimized for post-immunoprecipitation western blot detection of primary antibodies without interference from denatured IP antibody fragments.
|
|
|
|
|
|
|
|
|
|
Make sure that the caps are tightly locked to prevent evaporation of water.
|
|
|
|
|
|
|
|
|
|
|
|
|
Reaction Mixture for 1 sample is as follows: