Search Thermo Fisher Scientific
We offer purified and agarose-immobilized proteases for enzymatic proteolysis (cleavage or digestion) of proteins to facilitate amino acid sequencing, peptide analysis, and polypeptide structural characterization.
For protein digestion using mass spectrometry, we offer a wide range of highly purified and MS-validated proteases.
Enzyme | Purified protease | Immobilized proteases | Specificity |
---|---|---|---|
Factor Xa | 32521 32520 | N/A | Cleaves at the carboxyl side of Ile-Glu-Gly-Arg sequence |
Human Rhinovirus 3C Protease (HRV 3C) | 88946 | N/A | Highly specific for the recognition sequence Leu-Glu-Val-Leu-Phe-Gln-↓-Gly-Pro and cleaves after the glutamine residue |
S. aureus V-8 Protease | 20195 | 20151 | Cleaves at the carboxyl side of aspartate and glutamate residues |
Enzyme | Purified protease | Immobilized proteases | Specificity |
---|---|---|---|
Trypsin, MS Grade | 90057 90058 90059 90305 | N/A | Cleaves at the carboxyl side of arginine and lysine residues |
Lys-C Protease, MS Grade | 90051 | N/A | Cleaves at the carboxyl side of lysine residues |
Trypsin/Lys-C Protease Mix, MS Grade | A41007 | N/A | Cleaves at the carboxyl side of lysine residues |
Lys-N Protease, MS Grade | 90300 90301 | N/A | Cleaves at the amino side of lysine residues |
Asp-N Protease, MS Grade | 90053 | N/A | Cleaves at the amino side of aspartate residues |
Chymotrypsin, MS Grade | 90056 | N/A | Cleaves at the carboxyl side of tyrosine, phenylalanine, tryptophan, and leucine |
Glu-C Protease, MS Grade | 90054 | N/A | Cleaves at the carboxyl side of glutamate or aspartate |
Enzyme | Purified protease | Immobilized proteases | Specificity |
---|---|---|---|
Proteinase K | 17916 | N/A | Cleaves at the carboxyl side of aliphatic, aromatic, and hydrophobic residues |