Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Clinical personal protective equipment (PPE) is the key to a safer, healthier working environment. Our range has you and your patients covered from head to toe with lightweight and comfortable fabrics that meet national health standards.
These products are not available for purchase by the general public.
With the current climate we live in, self-testing is imperative in our everyday life. The Flowflex Nasal COVID-19 RAT Kit provides the convenience and ease needed for this task.
Learn moreThe Promed range of disposable cover/isolation gowns offers a range of styles and features to provide the right protection against hospital acquired infections for healthcare workers. This range of cover gowns has been designed to meet the ANSI/AAMI PB70 standards spanning from level 1-4.
Our broad range of non-sterile examination gloves provides the right protection for medical professionals, patients, and industry. Available in nitrile, latex, and vinyl options, we have a suitable product for all industry environments.
Our surgical and general face masks are available in a variety of styles, features, and levels of protection required to meet the (AS) 4381:2015 industry standard spanning from level 1–3.
Promed offers a variety of protective headwear that can be used in a wide range of applications across the healthcare sector.
Promed shoe covers are made from durable PP Spunbond coated PE. Providing superior grip, safety, and fluid resistance the shoe covers can be used in a wide range of applications for protection.