Antibodies
Beakers
Chemicals
Lab Centrifuges
Lab Equipment
Lab Plasticware and Supplies
PCR Equipment and Supplies
Pipettes and Pipette Tips
Ultra Low-Temperature Freezers
Spectroscopy
See all product categories
Applications
Bioprocessing
Clinical and Diagnostics
Digital Solutions
Environmental
Food and Beverage
Hospital and Patient Care
Industrial and Applied Sciences
Lab Solutions
Life Sciences
Manufacturing and Processing
See all applications and techniques
Services
Compliance, Validation and Calibration Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Service Plans
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
Home
›
Industrial & Applied Sciences
›
Chromatography
›
Chromatography Learning Center
›
HPLC Learning Center
›
HILIC HPLC and UHPLC Columns Information
›
HILIC Overview
›
Advantages of HILIC
Advantages of HILIC
See Navigation
‹
HILIC Overview
HILIC Stationary Phases
›
HILIC Retention Mechanisms
›
Advantages of HILIC
›
Achieve better separation efficiency under highly aqueous conditions
Retention of hydrophilic compounds that are difficult to retain by Reversed Phase Liquid Chromatography (RPLC)
Improved sensitivity with MS detection (due to more efficient desolvation of the analytes)
Compatibility with SPE extracts which are often in 100% organic solvent
Low back pressures due to the low viscosity of the organic-rich mobile phases
Separation efficiency of very polar analytes in HILIC better than in RPLC under highly aqueous mobile phase
Comparison chromatograms
Column:
Thermo Scientific™ Hypersil GOLD™, 5µm, 150 x 2.1mm
Mobile phase:
H2O/ MeCN (98:2) + 0.1% formic acid
Detection:
+ESI (spray conditions adjusted for higher aqueous content of mobile phase)
Injection:
1ng on column
Column:
Thermo Scientific™ Hypersil GOLD™ HILIC, 5µm, 150 x 2.1mm
Mobile phase:
Ammonium formate 50mM pH 3.5/ MeCN (10:90)
Detection:
+ESI Injection: 1ng on column
Related products
Zwitterionic HILIC Columns
PEI HILIC Columns
Silica HILIC Columns
Urea HILIC Columns
Amide HILIC Columns
CMD Wide-format style fixes
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
CMD SchemaApp code