Search Thermo Fisher Scientific
Achieve ultimate performance in automated pipetting with Thermo Scientific™ D.A.R.T.'S™. These tips offer a unique face seal design with a magazine rack format that helps result in optimal accuracy and precision. Choose from a variety of tip options to meet your pipetting needs.
Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x2, Matrix™ PlateMate™ 2x3, PlateMate™ Plus Systems and Versette.
![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
30µL | 300µL | 300µL Extended-Length | 300µL Extended-Length Wide-Bore | 30µL 384 | 100µL 384 |