The Thermo Scientific iFast Developer's Tool Kit combines the flexibility of scripting with the ease of use of graphical programming to make recipe creation faster and easier than ever before. 
The iFast Developer's Tool Kit enables operators to quickly and easily create new or modify existing recipes to fit their specific needs. Recipes are edited within a new intuitive graphical environment, enabling faster learning of the programming environment as well as an easier understanding of existing recipes. Within the iFast Developer's environment, the operator is able to control column settings, detector settings, stage motion and position, patterning and site alignments with logical operation and looping. This enables a high degree of customization to match specific application requirements. 
iFast Recorder is included with iFast Developer's Tool Kit allowing to record recipes interactively on an instrument both for unattended operation as well as the creation of a baseline recipe for further enhancements with the Developer's Tool Kit.
Key Features
  • Graphical programming environment.
  • Runner for unattended operation.
  • Alignment tools: image recognition and edge finding.
  • FIB, SEM, Stage, GIS, patterning and other controls.
  • Macro recorder for faster recipe creating.
  • Process looping.
  • Logical tests such as If-Then statements.
Applications
pathfinding_thumb_274x180_144dpi
Semiconductor Pathfinding and Research
Advanced electron microscopy, focused ion beam, and associated analytical techniques for identifying viable solutions and design methods for the fabrication of high-performance semiconductor devices.
yield_ramp_metrology_2_thumb_274x180
Yield Ramp and Metrology
We offer advanced analytical capabilities for defect analysis, metrology, and process control, designed to help increase productivity and improve yield across a range of semiconductor applications and devices.
Semiconductor Failure Analysis
Semiconductor Failure Analysis
Increasingly complex semiconductor device structures result in more places for failure-inducing defects to hide. Our next-generation workflows help you localize and characterize subtle electrical issues that affect yield, performance, and reliability.
physical_characterization_thumb_274x180_144dpi
Physical and Chemical Characterization
Ongoing consumer demand drives the creation of smaller, faster, and cheaper electronic devices. Their production relies on high-productivity instruments and workflows that image, analyze, and characterize a broad range of semiconductor and display devices.
Techniques
Style Sheet for Komodo Tabs
Semiconductor Analysis and Imaging
Thermo Fisher Scientific offers scanning electron microscopes for every function of a semiconductor lab, from general imaging tasks to advanced failure analysis techniques requiring precise voltage-contrast measurements.
Learn more ›
Semiconductor Analysis and Imaging
Thermo Fisher Scientific offers scanning electron microscopes for every function of a semiconductor lab, from general imaging tasks to advanced failure analysis techniques requiring precise voltage-contrast measurements.
Learn more ›
Style Sheet to change H2 style to p with em-h2-header class
Contact us
Electron microscopy services