Search Thermo Fisher Scientific
Crosslinkers contain reactive ends to specific functional groups, such as primary amines and sulfhydryls on proteins and antibodies, enabling the covalent joining of two or more biomolecules. Homobifunctional crosslinkers have the same reactive chemistry at both ends and heterobifunctional crosslinkers have different chemistries at each end. PEGylated crosslinkers are also available which provide enhanced solubility, increased stability, reduced aggregation, and reduced immunogenicity to proteins.
Use our Crosslinker Selection Tool to find the optimal protein crosslinker reagent for your protein function or interaction application, or access our helpful Bioconjugation Technical Handbook to help improve your crosslinking results.
Crosslinkers, also known as bifunctional crosslinkers, are reagents that contain two or more reactive groups which covalently attach via a spacer to functional groups on proteins or other biomolecules. Three types of crosslinkers are available: homobifunctional crosslinkers, heterobifunctional crosslinkers, and photoreactive crosslinkers. Homobifunctional crosslinking reagents have identical reactive groups, primarily amine-to-amine or sulfhydryl-to-sulfhydryl. They are typically used to form intramolecular crosslinks or to prepare polymers from monomers. Heterobifunctional crosslinking reagents have different reactive groups such as amine-to-sulfhydryl, carboxyl-to-amine, or sulfhydryl-to-carboxyl. They are useful for preparing conjugates between two different biomolecules. Heterobifunctional crosslinking reagents also include photoreactive crosslinking reagents that react with nucleophiles or form C-H insertion sites after exposure to UV light. Learn more about crosslinking reagents.
Find the optimal crosslinking reagent for your application using our Crosslinker Selection Tool or select from the most common crosslinkers below.
Sulfhydryl -to-Sulfhydryl Crosslinkers | Order product | Spacer Arm (Å) | Cleavable? | Water-soluble? | Membrane permeable? |
---|---|---|---|---|---|
NHS-Maleimide | SMCC | 8.3 | N/A | No | Yes |
Sulfo-SMCC | 8.3 | N/A | Yes | No | |
NHS-Pyridyldithiol Crosslinkers | SPDP | 6.8 | Thiol | No | Yes |
LC-SPDP | 15.7 | Thiol | No | Yes |
Reactive group | Order product | Spacer Arm (Å) | Cleavable? | Water-soluble? | Membrane permeable? |
---|---|---|---|---|---|
NHS-Maleimide | |||||
SMCC | 8.3 | N/A | No | Yes | |
Sulfo-SMCC | 8.3 | N/A | Yes | No | |
NHS-Pyridyldithiol Crosslinkers | SPDP | 6.8 | Thiol | No | Yes |
Sulfo-LC-SPDP | 15.7 | Thiol | Yes | No |
Heterobifunctional crosslinking.Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl) cyclohexane-1-carboxylate), an amine-to-sulfhydryl crosslinker, contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane-stabilized spacer arm. Heterobifunctional crosslinkers allow for sequential reactions with two molecules to control the conjugation without polymerization.
Reactive group | Order product | Spacer Arm (Å) | Cleavable by? | Water-soluble? | Membrane permeable? |
---|---|---|---|---|---|
NHS ester/ aryl azide | |||||
Sulfo-SANPAH | 18.2 Long | No | Yes | No | |
NHS ester/ diazirine | SDA | 3.9 Short | No | No | Yes |
Sulfo-SDA | 3.9 Short | No | Yes | No | |
LC-SDA | 12.5 Mid | No | No | Yes | |
Sulfo-LC-SDA | 12.5 Mid | No | Yes | No | |
SDAD | 13.5 Mid | Thiols | No | Yes | |
Sulfo-SDAD | 13.5 Mid | Thiols | Yes | No |
Heterobifunctional kits are offered to conjugate and purify a protein with a reactive maleimide or DBCO label for reaction with a sulfhydryl or azide, respectively.
Reactive group | Order product | Spacer Arm (Å) | Labeling scale | Number of reactions | Labeled protein reactive towards |
---|---|---|---|---|---|
TFP Ester/DBCO | DBCO Protein Labeling Kit | 17.9 | 0.1–0.5 mg | 5 | Azides |
NHS-Maleimide | Maleimide Protein Labeling Kit | 8.3 | 0.1–1 mg | 5 | Sulfhydryls |
Controlled Protein-Protein Crosslinking Kit | 11.6 2.8 | 0.5-5 mg | 1 | Amine Sulfhydryl |
* Not determined
Sulfhydryl -to-Sulfhydryl Crosslinkers | Order product | Spacer Arm (Å) | Cleavable? | Water-soluble? | Membrane permeable? |
---|---|---|---|---|---|
NHS-Maleimide | SMCC | 8.3 | N/A | No | Yes |
Sulfo-SMCC | 8.3 | N/A | Yes | No | |
NHS-Pyridyldithiol Crosslinkers | SPDP | 6.8 | Thiol | No | Yes |
LC-SPDP | 15.7 | Thiol | No | Yes |
Reactive group | Order product | Spacer Arm (Å) | Cleavable? | Water-soluble? | Membrane permeable? |
---|---|---|---|---|---|
NHS-Maleimide | |||||
SMCC | 8.3 | N/A | No | Yes | |
Sulfo-SMCC | 8.3 | N/A | Yes | No | |
NHS-Pyridyldithiol Crosslinkers | SPDP | 6.8 | Thiol | No | Yes |
Sulfo-LC-SPDP | 15.7 | Thiol | Yes | No |
Heterobifunctional crosslinking.Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl) cyclohexane-1-carboxylate), an amine-to-sulfhydryl crosslinker, contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane-stabilized spacer arm. Heterobifunctional crosslinkers allow for sequential reactions with two molecules to control the conjugation without polymerization.
Reactive group | Order product | Spacer Arm (Å) | Cleavable by? | Water-soluble? | Membrane permeable? |
---|---|---|---|---|---|
NHS ester/ aryl azide | |||||
Sulfo-SANPAH | 18.2 Long | No | Yes | No | |
NHS ester/ diazirine | SDA | 3.9 Short | No | No | Yes |
Sulfo-SDA | 3.9 Short | No | Yes | No | |
LC-SDA | 12.5 Mid | No | No | Yes | |
Sulfo-LC-SDA | 12.5 Mid | No | Yes | No | |
SDAD | 13.5 Mid | Thiols | No | Yes | |
Sulfo-SDAD | 13.5 Mid | Thiols | Yes | No |
Heterobifunctional kits are offered to conjugate and purify a protein with a reactive maleimide or DBCO label for reaction with a sulfhydryl or azide, respectively.
Reactive group | Order product | Spacer Arm (Å) | Labeling scale | Number of reactions | Labeled protein reactive towards |
---|---|---|---|---|---|
TFP Ester/DBCO | DBCO Protein Labeling Kit | 17.9 | 0.1–0.5 mg | 5 | Azides |
NHS-Maleimide | Maleimide Protein Labeling Kit | 8.3 | 0.1–1 mg | 5 | Sulfhydryls |
Controlled Protein-Protein Crosslinking Kit | 11.6 2.8 | 0.5-5 mg | 1 | Amine Sulfhydryl |
* Not determined
Protein crosslinking no-weigh packaging formats offer quick and convenient ready-to-use solutions. This format eliminates the need to weigh out small amounts of dry compounds and eliminates the waste of unused material. Simply reconstitute and the reagent is ready to use at the desired concentration.
Protein crosslinking premium grade reagents are high-quality reagents formulated for applications where product integrity and risk minimalization are essential. These premium grade reagents are ideal for long-term research projects that require consistent performance. Crosslinker premium grade products provide quality assurance review, lot sample retention, change control notification, along with an extra level of analytical testing and chemical characterization.
Test parameter | Test method | Premium grade | Standard grade |
---|---|---|---|
Purity | Quantitative NMR using an internal standard | Yes | Yes |
Visual | Color assessment | Yes | Yes |
Solubility | Sample dissolves at specified concentration in deionized water to yield a clear, colorless solution | Yes | Yes |
Identity | Infrared spectroscopy | Yes | Yes |
Mass identity | Mass spectrometry | Yes | NA |
Water content | Karl Fischer titration | Yes | NA |
Trace metals | Inductively coupled plasma mass spectrometry (ICP-MS) | Yes | NA |
Elemental analysis | Combustion analysis (values reported for C, H, N, O, and S) | Yes | NA |
Residual solvent analysis | Headspace gas chromatography | Upon request | NA |
In vivo crosslinking can be used to characterize protein interactions and ligand-receptor interactions irrespective of treatment conditions. Various sizes of in vivo crosslinkers are available to target surface and intracellular proteins for analysis by different methods such as immunoprecipitation (IP), Co-IP, chromatin immunoprecipitation (ChIP), electrophoresis mobility shift assay (EMSA), western blot, immunofluorescence (IF), and immunohistochemistry (IHC).
NHS-ester crosslinkers are widely used for in vivo crosslinking. NHS-ester, amine-specific reactions are fast and highly efficient since lysine residues are abundant and easily accessible on the surface of most proteins. Homobifunctional crosslinkers use a “shotgun” type approach for identifying protein complexes since they crosslink all interacting molecules whose lysine residues are within the crosslinkers spacer length. Subsequent detection simply requires a specific antibody or probe that targets one of the molecules in the complex.
Find homobifunctional, amine-reactive, NHS-ester crosslinkers for in vivo crosslinking
NHS-ester/diazirine crosslinkers combine NHS-ester chemistry with diazirine-based photochemistry to create crosslinking. While typical heterobifunctional crosslinkers require specific amino acid groups at correct distances, SDA crosslinkers utilize a “two-step” reaction by crosslinking one protein using the NHS-ester then UV light to activate crosslinking via the diazirine moiety to any amino acid on a second protein.
Find heterobifunctional, photoreactive, NHS-ester/diazirine crosslinkers for in vivo crosslinking
Photo-Leucine and Photo-Methionine, used in metabolic labeling, are incorporated into proteins by protein synthesis (3). These non-toxic photo-amine acids are non-invasive and do not alter the cell’s metabolism. Photo-Leucine and Photo-Methionine are highly specific and ideal for studying hydrophobic interactions. Further, they can be used for either extracellular or intracellular crosslinking.
Find photoreactive amino acid crosslinkers for in vivo crosslinking
Everything you need to bioconjugate, crosslink, biotinylate, and modify proteins and peptides.
Need larger quantities of our protein labeling, crosslinking or modification reagents? We offer bulk and custom fill sizes.