Data Acquisition and Signal Analysis
Browse data acquisition systems
Solutions for measurement and analysis of sound events
HEAD Acoustics has developed hardware and software solutions for measuring and analysing sound events on the basis of aurally-accurate recording and playback systems, which have become today's industry standard. We offer a range of frontends for data acquisition and signal analysis including handheld models for portable applications, as well as modular systems that support diverse sensor types.
Handheld data acquisition systems
SQuadriga III 
SQuadriga III compact hand-held system with its 8 analog channels and various digital inputs is ideal for mobile sound and vibration measurements in any industrial application.
SQobold 
SQobold is a compact and portable 4-channel data acquisition system and supports CAN FD/OBD-II - to help with automotive characteristics such as vehicle speed, and engine RPM.
Accessories
Modular and scalable data acquisition systems
HEADlab II 
Modular controllers with various input, playback and power supply modules. Clear and rugged design for multi-channel sound and vibration measurement. HEADlab II systems provide high data rates, CAN-FD support and synchronization capabilities via GPS and LAN (PTP).
HEADlab modules 
Individual, easy to configure digital and analog modules supporting diverse sensor types, that users choose which modules they want to combine with a controller.
The following modules are available:
Controllers and digital input
  • labCTRL II.1: Second generation HEADlab controller for data concentration and synchronization with HEADlink 2.0
  • labDX: Digital input module with CAN FD, FlexRay, HMS and pulse interfaces
  • labHRT6: Digital input module for high-resolution measurement of speeds
Analog modules
  • labM6 II: 6-channel voltage/microphone input module of the 2nd HEADlab genera­tion with HD wide-range input (High Dynamics)
  • labVF6 II: 6-channel voltage/ICP® input module of the 2nd HEADlab generation with switchable low-pass filters
  • labV12 II: 12-channel voltage/ICP® input module of the 2nd HEADlab generation for applications with larger numbers of channels
  • labV24 II: 24-channel voltage/ICP® input module of the 2nd HEADlab generation for applications with high channel numbers
  • SCU I6: 6-channel adapter to perform current measurements with HEADlab modules and compact systems
  • labV6HD: 6-channel line/ICP® input module with wide-range input (high dynamics)
  • labSG6: 6-channel input module for connecting resistive measuring bridges (strain gages) and more
  • labCF6: 6-channel charge/ICP® input module with integrated charging amplifiers
  • labT6: 6-channel input module for connecting thermal elements of type K or RTD (PT100, PT1000)
  • labO2 – 2-channel playback equalizer with line outputs
  • labHSU – High-end dual-channel data acquisition module
Power supply
  • labPWR I.1: HEADlab Power-Box for supplying power to individual modules or smaller systems up to 40 W
  • labPWR I.2: HEADlab Power-Box for supplying power to larger systems up to 100 W
  • labPWR I.3: HEADlab Power-Box for supplying labHSU, HEADlab systems up to 35 W and SQuadriga III
Accessories
A wide range of accessories are available, contact our experts to discuss your requirements.
labHSU dual-channel module
The labHSU can be used as a frontend, as a module in a HEADlab system or as a stand-alone system by remote control via smartphone, tablet or the RC X remote. The labHSU headphone output offers standard equalization for various headphone types which enables aurally accurate monitoring of a recording directly on the device.
labCOMPACT12 and labCOMPACT24
labCOMPACT is a compact, simplified HEADlab system bringing together the basic functions of a controller with a multi-channel module. This module is ideal for multi-channel analysis in acoustic optimisation of household appliance and office equipment.
labCORE is the modular, all-in-one solution for high-precision voice and audio quality testing and tailored for the use the ACQUA software. Users can adapt the hardware platform exactly to individual test scenarios and flexibly expand it at any time as required. Applications include mobile phones, headphones, IoT devices like smart speakers or in-vehicle hands-free devices and ICC systems.
Flexibly expand the labCORE hardware platform to adapt it to your specific measurement purposes. This hardware platform is ideally suited for testing voice and audio quality of devices in various fields of applications and with different transmissions technologies.
The following extensions are available:
Analog modules
Digital and interfacing
Software extensions
  • coreBEQ (Code 7740), Binaural equalization
  • coreBEQ-Add (Code 7741), Additional binaural equalization
  • coreIP (Code 7770), Voice over IP extension
  • coreIP-IMP (Code 7771), VoIP impairment extension
  • coreIP-EVS (Code 7773), EVS extension
  • coreIP-OPUS (Code 7774), OPUS extension
  • coreIP-Alexa (Code 7775), Enables the hardware platform to act as an Amazon Alexa reference client
HIB I (Headset Interface Box) is used for voice and audio quality testing of headphones, headsets and the headset functionality of telephones, by serving as a reference interface.
HAC III is a probe coil combined with a handset positioner. The positioner has a large mounting area which enables it to accomodate all handset sizes. The probe coil then can verify the hearing aid compatibility of telephony handsets by measuring their magnetic coupling capabilities.
HPO is a lightweight and compact hardware to simulate a Plain Old Telephone System (POTS) terminal in order to test the POTS functionality of IP-based home gateways, DSL and cable modems as well as other devices with a POTS interface. 
Directional testing
HRT I
HRT I is ideally suited for fully automated orientation-dependent acoustic tests, and the rotation unit of the turntable offers a 360° rotation range, which can be approached in 0.1° steps. The turntable rotates all kinds of test objects including telephony devices, video conferencing systems, loudspeakers, microphones or measurement equipment.
HRR I
HRR I is a motorised rotating reflector to influence the acoustic coupling between loudspeaker and microphones during acoustic tests, with users able to quickly change the acoustic echo path. For instance, in hands-free communication testing, identify and then optimise the system’s general communication quality and echo attenuation performance in a fully automated and repeatable way.
Analysis software
We offer comprehensive software solutions from HEAD Acoustics, and due to the modular structure of these software platforms, users can tailor specific solutions to their individual requirements. 
  • Sound and vibration analysis
  • Telecommunication and audio analysis
Discuss your needs with an expert
Speak to our technical experts to discuss your application requirements