Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Coriolis µ is an innovative biological air sampler for bio-contamination assessment, dedicated to air quality control and air quality monitoring in environmental and pollution research, the pharmaceutical, food, and veterinary industries, and in biomedical and health environments.
Based on a cyclonic technology combined with a high air flow rate, Coriolis μ offers the most efficient particle collection in 10 minutes. Biological particles such as toxins, viruses, bacteria, moulds, pollens, and spores are collected and concentrated in a liquid, ready to be analysed with a range of downstream experiments and biological analysis methods such as culturing, immunological tests, next generation sequencing, spectrometry, and more.
Extend monitoring time up to 6 hours by automatically maintaining a constant liquid collection volume with the Long Time Monitoring option. This also comes with an adjustable tripod stand for easy deployment in environmental monitoring applications.
Catalog number | Description | |
---|---|---|
P001080-CORM0-A.0 | Bertin Coriolis Micro air sampler | Request quote › |
S001097-CORM0-A.0 | Bertin Long Time Monitoring option for Coriolis Micro | Request quote › |
P002055-CORC0-A.0 | Bertin Coriolis Compact air sampler | Request quote › |