Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
IUL Spin Air samplers are a compact, portable, simple solution to air sampling and microbial air contamination monitoring.
The IUL Spin Air sampler is a very compact unit designed for easy transportation and sampling anywhere needed.
The system’s highlights include:
Spin Air | Spin Air (Secondary) | Spin Air Basic | |
---|---|---|---|
![]() | ![]() | ![]() | |
RS 232 connection (PC/Printer, XML, CSV) | * | ||
Incorrect airflow alarm–abort function | * | ||
LIMS connectivity | * | ||
Bar code connection (IUL bar code device) | * | ||
Simultaneous sampling (connectivity to 2nd unit) | * | ||
Supplied with 90 mm plate holder and cover | Aluminium | Aluminium | Aluminium |
Weight | 1.7 kg | 1.3 kg | 1.35 kg |
* Through primary unit
Catalog number | Description | |
---|---|---|
IUL5500 | Spin Air Sampler | Request quote › |
IUL5502 | Spin Air Secondary Sampler | Request quote › |
IUL5532 | Spin Air Basic Sampler | Request quote › |