Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The 3M™ Clean-Trace™ Surface Protein Plus Test Swab PRO100 can be integrated into any Hygiene Management Program so risk areas can be monitored for cleaning effectiveness.
The 3M Clean-Trace Surface Protein Plus Test Swab PRO100 is a simple and reliable method of checking for protein residues after cleaning and identifying contamination.
This sensitive tool detects as little as 50 µg of protein on surfaces. A quick, easy visual reading of a color change indicates the presence of detectable levels of protein, and fast results allow immediate corrective action.
The system helps enable you to:
![]() | ![]() | ![]() | ![]() |
1. Swab. surface firmly. If the surface is dry, wet the swab with the moisturiser supplied in the kit. | 2. Click. Activate the swab, and shake from side to side for 5 seconds. | 3. Read. Leave the swab upright at room temperature for 10 minutes. Read the result. | 4. Interpret results. |
Catalog number | Description | Quantity | |
---|---|---|---|
3MPR0100 | Clean-Trace Surface Protein Plus Swabs | 100 pcs | Request quote › |