Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The Neogen AccuPoint Advanced ATP system provides customers assurance by producing consistent and reliable data for evaluating sanitation program effectiveness in food processing and food services facilities.
ATP (adenosine triphosphate) is a chemical compound found in all living cells, including bacteria, food debris, yeast, and mould. The Neogen Accupoint Advanced ATP systems utilizes ATP bioluminescence to determine the cleanliness of test samples.
The system’s highlights include:
Catalog number | Description | |
---|---|---|
NEO9905 | AccuPoint Advanced Surface Sampler | Request quote |
NEO9907 | AccuPoint Advanced Access Sampler | Request quote |
NEO9906 | AccuPoint Advanced Water Sampler | Request quote |
NEO9603 | AccuPoint Advanced RFID Signs | Request quote |
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.