Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The IUL DOT Colony Counter is an easy and inexpensive system to review, read, and count colonies on a variety of agar plates.
The IUL DOT Colony Counter allows agar plates to be inserted inside the colony counter where they are properly lit and magnified. Operators can tick detected colonies with a specifically devised marker while a digital display increases the total counts. Pointers that can directly contact colonies are also available.
The system’s highlights include:
Catalog number | Description | |
---|---|---|
IUL90008700 | DOT Colony Counter with 120 mm Magnifying Glass with Light | Request quote › |
IUL90008701 | DOT Colony Counter DOT System with 100 mm Magnifying Glass | Request quote › |
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.