Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The IUL Petri Labeler automatically tags pre-printed barcode labels on the side walls of Petri plates. Operating the unit is as simple as feeding a stack of Petri plates on one end, pushing the start button, and collecting labeled plates at the other end.