Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Activation of caspase enzymes, which participate in the cleavage of protein substrates and in the subsequent disassembly of the cell, plays an important role in apoptosis. Thermo Fisher Scientific offers reagents and kits for imaging caspase activity in real-time or as an end-point assay.
Caspases are a family of cysteine–aspartic acid specific proteases (CED-3/ICE) that have been identified as crucial mediators of the complex biochemical events associated with apoptosis (1). Caspases are divided into two groups: initiator and effector caspases, and once activated by the initiator caspase (e.g. caspase-8 and -9), the effector caspases (e.g. caspases-3 and -7) function to cleave protein substrates and initiate apoptosis (2). The recognition site for each caspase is marked by three to four amino acids followed by an aspartic acid residue, with the cleavage occurring after the aspartate. The caspase proteases are typically synthesized as inactive precursors and inhibitor release or cofactor binding activates the caspase through cleavage at internal aspartates, either by autocatalysis or by the action of another protease.
Not all caspases are involved with apoptosis and in recent years, it has been demonstrated that some caspases may be involved in inflammation (3). Because cell death cascades are complex and dynamic, it is highly recommended that a multi-parametric approach is conducted for an accurate assessment of apoptosis.
CellEvent Caspase-3/7 detection reagent represents a new generation of caspase substrates and is an important tool for the study of apoptosis. It is available in both green (Ex/Em: 502/530) and red (Ex/Em: 590/610) and can be visualized with a FITC or Texas Red filter (respectively) on any fluorescence microscope. The cell-permeant CellEvent reagent comprises the four–amino acid peptide DEVD—which contains the recognition site for caspases 3 and 7—conjugated to a nucleic acid–binding dye. Because the DEVD peptide inhibits the ability of the dye to bind to DNA, CellEvent Caspase-3/7 Green detection reagent is intrinsically nonfluorescent. In the presence of activated caspase 3/7, the dye is cleaved from the DEVD peptide and free to bind DNA, producing a bright fluorescent signal indicative of apoptosis (Figure 1). This robust assay is highly specific for caspase 3/7 activation and, as expected, we observe nearly complete inhibition of the CellEvent Caspase-3/7 Green detection reagent signal in cells pretreated with a caspase 3/7 inhibitor (Figure 2).
One important advantage of the CellEvent caspase-3/7 assay is that no wash steps are required, thus preserving fragile apoptotic cells that are typically lost during these rinses. The loss of apoptotic cells during wash steps may lead to an underestimation of the extent of apoptosis in the sample, resulting in poor assay accuracy. Additionally, the fluorescent signal resulting from cleavage of CellEvent Caspase-3/7 detection reagent survives formaldehyde fixation and detergent permeabilization, providing the flexibility to perform endpoint assays and to probe for other proteins using immunocytochemical techniques. The newest Caspase-3/7 reagent, CellEvent Caspase-3/7 Red, can also be imaged easily alongside GFP expression in live cells with little to no cross talk (Figure 3).
TMRM and CellEvent Caspase-3/7 Green used to monitor apoptosis progression in HeLa cells treated with staurosporine. HeLa cells were loaded with 50 nM TMRM (red) followed by 5 µM CellEvent Caspase 3/7 Reagent (green). Cells were then treated with 0.5 µM staurosporine and images were acquired every 5 minutes over 7 hours on a Molecular Devices ImageXpress at 10x. Over the 7 hour time course, staurosporine induced a loss of mitochondrial membrane potential followed by activation of caspase 3/7 as evidenced by a decrease in TMRM signal (red) and an increase in caspase 3/7 signal (green), respectively.
Figure 1. No-wash apoptosis detection in live cells. HeLa cells were loaded with 7.5 μM CellEvent Caspase-3/7 Green detection reagent and treated with vehicle control (A) or 0.5 μM staurosporine (B) for 4 hr. Untreated cells were also stained with Hoechst 33342 (blue) to visualize the nuclei of negative cells. The fluorescence signal is overlaid with differential interference contrast (DIC). Cells treated with staurosporine show a significant increase in signal from CellEvent Caspase-3/7 Green detection reagent (green).
Figure 2. Inhibition of caspase 3/7 activity with a caspase inhibitor. HeLa cells were treated with 0.5 μM staurosporine in the presence of 0–30 μM Caspase 3/7 Inhibitor 1 (EMD Chemicals) for 4 hr. Cells were then labeled with 5 μM CellEvent Caspase-3/7 Green detection reagent and Hoechst 33342 (blue) in complete medium. (A) In a dose-dependent manner, the Caspase 3/7 Inhibitor 1 decreased the signal from CellEvent Caspase-3/7 Green detection reagent in staurosporine-treated cells, expressed quantitatively as percent positive cells. (B) Visualization of staurosporine-induced caspase 3/7 activity (green); and (C) inhibition thereof is also presented. Images were acquired and analyzed on the Thermo Scientific Cellomics ArrayScan VTI platform.
Figure 3. Expression of GFP with Caspase Red shows minimal crosstalk. A673 and U2OS cells were plated at 3,000 cells per well and transduced with cytosolic GFP BacMam and treated with DMSO or 2 µM camptothecin for 24 hours at 37°C. Cells were subsequently stained with CellEvent Caspase 3/7 Red for 1 hour at 37°C prior to imaging on an EVOS M7000 widefield fluorescence microscope.
The Image-iT LIVE Caspase Detection Kits employ a novel approach to detect active caspases that is based on a fluorescent inhibitor of caspases (FLICA) methodology, essentially an affinity label. The reagent associates a fluoromethyl ketone (FMK) moiety, which can react covalently with a cysteine, with a caspase-specific amino acid sequence. For the caspase-3 and -7 reagents, this recognition sequence is aspartic acid-glutamic acid- valine-aspartic acid (DEVD); for the poly caspases reagents, this recognition sequence is valine-alanine-aspartic acid (VAD) and for caspase-8 reagent this recognition sequence is leucine-glutamic acid-threonine-aspartic acid (LETD). A fluorescent dye is attached as a reporter. The FLICA reagent is thought to interact with the enzymatic reactive center of an activated caspase via the recognition sequence, and then to attach covalently through the FMK moiety. The FLICA inhibitor is cell permeant and noncytotoxic. Unbound FLICA molecules diffuse out of the cell and are washed away; the remaining fluorescent signal is a direct measure of the amount of active caspase that was present at the time the inhibitor was added (Figure 4). Each kit contains a cell-impermeant nucleic acid stain to monitor plasma membrane integrity.
FLICA reagents have been used widely to study apoptosis using flow cytometry and microscopy (4-8). However it should be noted that while cellular fluorescence from the reagent is strongly linked to caspase activity in apoptotic cells, its interaction with other cellular sites may contribute to signal intensity in non-apoptotic cells (9). Appropriate controls should be included in any experimental design.
Figure 4. Live HeLa cells visualized using the Image-iT LIVE Red Poly Caspase Detection Kit. Live HeLa cells visualized using the Image-iT LIVE Red Poly Caspase Detection Kit (I35101). Cells were treated with staurosporine to induce apoptosis, and FLICA reagent (SR-VAD-FMK, left) was added. Cells were then loaded with SYTOX Green dye (middle) and Hoechst 33342 (blue fluorescence, right) and analyzed by fluorescence microscopy. Apoptotic, non-apoptotic, and dead cells are all visible in the same field.
CellEvent Caspase-3/7 Green Detection Reagent | CellEvent Caspase-3/7 Red Detection Reagent | Image-iT LIVE Caspase-3 and -7 Detection Kits | |||||
---|---|---|---|---|---|---|---|
Application | No-wash assay to monitor ongoing caspase-3 and -7 activity. | End point assay for the detection of active caspase-3 and -7. | |||||
Readout | Following interaction with active caspase-3 and -7 in apoptotic cells, the fluorogenic, cell-permeant substrate is able to bind DNA and fluoresce brightly. | Fluorescent inhibitor of caspase-3 and -7 interacts with the caspase enzymatic site. Following a wash step, the remaining fluorescence signal is a direct measure of the amount of active caspase at the time the inhibitor was added. | |||||
Caspase target and function | Caspases-3 and -7 are downstream effector caspases that, when activated, cleave protein substrates and execute apoptosis. | ||||||
Detection reagent | DEVD conjugated to a nucleic acid binding dye | FAM-DEVD-FMK | SR-DEVD-FMK | ||||
Ex/Em (nm) | 502/530 | 590/610 | 488/530 | 550/595 | |||
Common filter | FITC | Texas Red | FITC | TRITC | |||
Nuclear counterstains | None included | None included |
|
| |||
Platform compatibility |
|
| |||||
Sample compatibility |
|
| |||||
Format | 1 vial (lyophilized powder)* | 5 vials (lyophilized powder)** | Dropper bottle (1.75ml, 12x in PBS) | 1 vial (lyophilized powder)* | 5 vials (lyophilized powder) | 25 assays | 25 assays |
Cat. No. | C10430 | C10431 | R37111 | C10430 | C10431 | I35106 | I35102 |
*1 vial=100 assays using 96-well plate; **5 vials=500 assays using 96-well plate |
Image-iT LIVE Caspase-8 Detection Kit | |
---|---|
Application | End point assay for the detection of active caspase-8. |
Readout | Fluorescent inhibitor of caspases interacts with the caspase-8 enzymatic site. Following a wash step, the remaining fluorescence signal is a direct measure of the amount of active caspase at the time the inhibitor was added. |
Caspase target and function | Caspase-8 is an initiator caspase. It functions to activate effector caspases (e.g. caspases-3 and -7). |
Detection reagent | FAM-LETD-FMK |
Ex/Em (nm) | 488/530 |
Common filter | FITC |
Nuclear counterstains |
|
Platform compatibility |
|
Sample compatibility |
|
Format | 25 assays |
Cat. No. | I35105 |
Image-iT LIVE Poly Caspases Detection Kits | ||
---|---|---|
Application | End point assay for the detection of active caspases. | |
Readout | Fluorescent inhibitor of caspases interacts with the caspase enzymatic site. Following a wash step, the remaining fluorescence signal is a direct measure of the amount of active caspase at the time the inhibitor was added. | |
Caspase target and function | Reagent reacts with multiple caspases (poly-caspases) that bind to the common recognition sequence valine-alanine-aspartic acid (VAD). | |
Detection reagent | FAM-VAD-FMK | SR-VAD-FMK |
Ex/Em (nm) | 488/530 | 550/595 |
Common filter | FITC | TRITC |
Nuclear counterstains |
|
|
Platform compatibility |
| |
Sample compatibility |
| |
Format | 25 assays | 25 assays |
Cat. No. | I35104 | I35101 |
CellEvent Caspase-3/7 Green Detection Reagent | CellEvent Caspase-3/7 Red Detection Reagent | Image-iT LIVE Caspase-3 and -7 Detection Kits | |||||
---|---|---|---|---|---|---|---|
Application | No-wash assay to monitor ongoing caspase-3 and -7 activity. | End point assay for the detection of active caspase-3 and -7. | |||||
Readout | Following interaction with active caspase-3 and -7 in apoptotic cells, the fluorogenic, cell-permeant substrate is able to bind DNA and fluoresce brightly. | Fluorescent inhibitor of caspase-3 and -7 interacts with the caspase enzymatic site. Following a wash step, the remaining fluorescence signal is a direct measure of the amount of active caspase at the time the inhibitor was added. | |||||
Caspase target and function | Caspases-3 and -7 are downstream effector caspases that, when activated, cleave protein substrates and execute apoptosis. | ||||||
Detection reagent | DEVD conjugated to a nucleic acid binding dye | FAM-DEVD-FMK | SR-DEVD-FMK | ||||
Ex/Em (nm) | 502/530 | 590/610 | 488/530 | 550/595 | |||
Common filter | FITC | Texas Red | FITC | TRITC | |||
Nuclear counterstains | None included | None included |
|
| |||
Platform compatibility |
|
| |||||
Sample compatibility |
|
| |||||
Format | 1 vial (lyophilized powder)* | 5 vials (lyophilized powder)** | Dropper bottle (1.75ml, 12x in PBS) | 1 vial (lyophilized powder)* | 5 vials (lyophilized powder) | 25 assays | 25 assays |
Cat. No. | C10430 | C10431 | R37111 | C10430 | C10431 | I35106 | I35102 |
*1 vial=100 assays using 96-well plate; **5 vials=500 assays using 96-well plate |
Image-iT LIVE Caspase-8 Detection Kit | |
---|---|
Application | End point assay for the detection of active caspase-8. |
Readout | Fluorescent inhibitor of caspases interacts with the caspase-8 enzymatic site. Following a wash step, the remaining fluorescence signal is a direct measure of the amount of active caspase at the time the inhibitor was added. |
Caspase target and function | Caspase-8 is an initiator caspase. It functions to activate effector caspases (e.g. caspases-3 and -7). |
Detection reagent | FAM-LETD-FMK |
Ex/Em (nm) | 488/530 |
Common filter | FITC |
Nuclear counterstains |
|
Platform compatibility |
|
Sample compatibility |
|
Format | 25 assays |
Cat. No. | I35105 |
Image-iT LIVE Poly Caspases Detection Kits | ||
---|---|---|
Application | End point assay for the detection of active caspases. | |
Readout | Fluorescent inhibitor of caspases interacts with the caspase enzymatic site. Following a wash step, the remaining fluorescence signal is a direct measure of the amount of active caspase at the time the inhibitor was added. | |
Caspase target and function | Reagent reacts with multiple caspases (poly-caspases) that bind to the common recognition sequence valine-alanine-aspartic acid (VAD). | |
Detection reagent | FAM-VAD-FMK | SR-VAD-FMK |
Ex/Em (nm) | 488/530 | 550/595 |
Common filter | FITC | TRITC |
Nuclear counterstains |
|
|
Platform compatibility |
| |
Sample compatibility |
| |
Format | 25 assays | 25 assays |
Cat. No. | I35104 | I35101 |
Fluorescence SpectraViewer
Online tool for visualization of the excitation and emission of fluorescent reagents. Tool allows for checking spectral compatibility for multiple fluorophores.
For Research Use Only. Not for use in diagnostic procedures.