Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Sartorius Entris II and Practum balances are compact, rugged, economically priced and suited for common day-to-day weighing procedures, perfect for the laboratory, training and school use. Models are available with readability from 1 g to 0.1 mg, and weight ranges up to 8.2 kg.
Every Sartorius balance offers quality, value and consistency. No matter what you’re weighing, the new Entris II is always the right choice. Offering unrivalled value at a budget price, this is the only balance in its class featuring isoCAL, LED touch technology and 12 built-in applications. Backed by almost 150 years of German engineering expertise and available in 40 different models, you’ll easily find an Entris II balance that meets your specific weighing needs.
Sartorius’s Entris II Essential line is the only balance in its class offering isoCAL, LED touch technology, and 12 built-in applications at a budget price. With over 40 models from which to choose, you will surely find a model to meet your basic weighing needs.
Highest quality standards
Easy clean for operational efficiency and durability
Effective draft shield
Adjustment | Internal and external |
Anti-theft locking device | Kensington lock, lockdown capability for cable or chain |
Application programs |
|
Levelling | Manual |
Linearity | 0.2 - 1000 mg (depending on model) |
Operating conditions - temperature | +10°C to +30°C |
Power requirements | 100 - 240 VAC, 50 - 60 Hz |
Repeatability | 0.08 mg - 500 mg (depending on model) |
Typical stabilisation time | ≤0.9 s - 1.5 s (depending on model) |
Under balance weighing | Yes |
Weighing chamber height | 240 mm (depending on model) |
Weighing units | Gram, kilogram, carat, pound, ounce, troy ounce, Hong Kong tael, Singapore tael, Taiwan tael, grain, pennyweights, milligram, parts per pound, China tael, mommes, Austrian carat, tola, baht, mesghal and Newton |
Display | LED screen with touch technology (graphic touch display on advance line models) |
Maximum weighing capacity | 60 g - 12,200 g (depending on model) |
Readability | 0.1 mg - 1000 mg (depending on model) |
Weighing pan dimensions | ⌀ 90, ⌀120 or 182 x 182 mm (depending on model) |
Data interface |
|
Sartorius’s Entris II Advanced line gives you additional value to the Entris II portfolio, with over 38 models offering benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display and 13 built-in application programs.
Added value for the Entris II Advanced line:
Adjustment | Internal and external |
Anti-theft locking device | Kensington lock, lockdown capability for cable or chain |
Application programs |
|
Levelling | Manual |
Linearity | 0.2 - 1000 mg (depending on model) |
Operating conditions - temperature | +10°C to +30°C |
Power requirements | 100 - 240 VAC, 50 - 60 Hz |
Repeatability | 0.08 mg - 500 mg (depending on model) |
Typical stabilisation time | ≤0.9 s - 1.5 s (depending on model) |
Under balance weighing | Yes |
Weighing chamber height | 240 mm (depending on model) |
Weighing units | Gram, kilogram, carat, pound, ounce, troy ounce, Hong Kong tael, Singapore tael, Taiwan tael, grain, pennyweights, milligram, parts per pound, China tael, mommes, Austrian carat, tola, baht, mesghal and Newton |
Display | LED screen with touch technology (graphic touch display on advance line models) |
Maximum weighing capacity | 60 g - 8200 g (depending on model) |
Readability | 0.1 mg - 100 mg (depending on model) |
Weighing pan dimensions | ⌀ 90, ⌀120 or 182 x 182 mm (depending on model) |
Data interface |
|
Get value for money without compromises in precision and reliability with the new Practum. Rely on consistent readings and excellent repeatability ensured by Sartorius quality and technology developed and designed in Germany. Practum has everything you need for standard applications.
Adjustment | With external weights |
Anti-theft locking device | Kensington lock, lockdown capability for cable or chain |
Application programs |
|
Levelling | Manual |
Linearity | 0.2 - 1000 mg (depending on model) |
Operating conditions - temperature | +10°C to +30°C |
Power requirements | 100 - 240 VAC, 50 - 60 Hz |
Repeatability | 0.1 mg - 500 mg (depending on model) |
Typical stabilisation time | ≤1 s - 2.0 s (depending on model) |
Underfloor weighing | Yes |
Weighing chamber height | 209 mm (depending on model) |
Weighing units | Gram, kilogram, carat, pound, ounce, troy ounce, Hong Kong tael, Singapore tael, Taiwan tael, grain, pennyweights, milligram, parts per pound, China tael, mommes, karat, tola, baht, mesghal and Newton |
Display | LED screen with touch technology |
Maximum weighing capacity | 0.1 mg - 500 mg (depending on model) |
Readability | 0.1 mg - 1000 mg (depending on model) |
Weighing pan dimensions | ⌀ 90 - 180 mm (depending on model) |
Data interface |
|
Sartorius’s Entris II Essential line is the only balance in its class offering isoCAL, LED touch technology, and 12 built-in applications at a budget price. With over 40 models from which to choose, you will surely find a model to meet your basic weighing needs.
Highest quality standards
Easy clean for operational efficiency and durability
Effective draft shield
Adjustment | Internal and external |
Anti-theft locking device | Kensington lock, lockdown capability for cable or chain |
Application programs |
|
Levelling | Manual |
Linearity | 0.2 - 1000 mg (depending on model) |
Operating conditions - temperature | +10°C to +30°C |
Power requirements | 100 - 240 VAC, 50 - 60 Hz |
Repeatability | 0.08 mg - 500 mg (depending on model) |
Typical stabilisation time | ≤0.9 s - 1.5 s (depending on model) |
Under balance weighing | Yes |
Weighing chamber height | 240 mm (depending on model) |
Weighing units | Gram, kilogram, carat, pound, ounce, troy ounce, Hong Kong tael, Singapore tael, Taiwan tael, grain, pennyweights, milligram, parts per pound, China tael, mommes, Austrian carat, tola, baht, mesghal and Newton |
Display | LED screen with touch technology (graphic touch display on advance line models) |
Maximum weighing capacity | 60 g - 12,200 g (depending on model) |
Readability | 0.1 mg - 1000 mg (depending on model) |
Weighing pan dimensions | ⌀ 90, ⌀120 or 182 x 182 mm (depending on model) |
Data interface |
|
Sartorius’s Entris II Advanced line gives you additional value to the Entris II portfolio, with over 38 models offering benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display and 13 built-in application programs.
Added value for the Entris II Advanced line:
Adjustment | Internal and external |
Anti-theft locking device | Kensington lock, lockdown capability for cable or chain |
Application programs |
|
Levelling | Manual |
Linearity | 0.2 - 1000 mg (depending on model) |
Operating conditions - temperature | +10°C to +30°C |
Power requirements | 100 - 240 VAC, 50 - 60 Hz |
Repeatability | 0.08 mg - 500 mg (depending on model) |
Typical stabilisation time | ≤0.9 s - 1.5 s (depending on model) |
Under balance weighing | Yes |
Weighing chamber height | 240 mm (depending on model) |
Weighing units | Gram, kilogram, carat, pound, ounce, troy ounce, Hong Kong tael, Singapore tael, Taiwan tael, grain, pennyweights, milligram, parts per pound, China tael, mommes, Austrian carat, tola, baht, mesghal and Newton |
Display | LED screen with touch technology (graphic touch display on advance line models) |
Maximum weighing capacity | 60 g - 8200 g (depending on model) |
Readability | 0.1 mg - 100 mg (depending on model) |
Weighing pan dimensions | ⌀ 90, ⌀120 or 182 x 182 mm (depending on model) |
Data interface |
|
Get value for money without compromises in precision and reliability with the new Practum. Rely on consistent readings and excellent repeatability ensured by Sartorius quality and technology developed and designed in Germany. Practum has everything you need for standard applications.
Adjustment | With external weights |
Anti-theft locking device | Kensington lock, lockdown capability for cable or chain |
Application programs |
|
Levelling | Manual |
Linearity | 0.2 - 1000 mg (depending on model) |
Operating conditions - temperature | +10°C to +30°C |
Power requirements | 100 - 240 VAC, 50 - 60 Hz |
Repeatability | 0.1 mg - 500 mg (depending on model) |
Typical stabilisation time | ≤1 s - 2.0 s (depending on model) |
Underfloor weighing | Yes |
Weighing chamber height | 209 mm (depending on model) |
Weighing units | Gram, kilogram, carat, pound, ounce, troy ounce, Hong Kong tael, Singapore tael, Taiwan tael, grain, pennyweights, milligram, parts per pound, China tael, mommes, karat, tola, baht, mesghal and Newton |
Display | LED screen with touch technology |
Maximum weighing capacity | 0.1 mg - 500 mg (depending on model) |
Readability | 0.1 mg - 1000 mg (depending on model) |
Weighing pan dimensions | ⌀ 90 - 180 mm (depending on model) |
Data interface |
|