Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
We offer a range of equipment and systems featuring weighing, measurement, and automation technology for laboratory and industrial applications. These ultra-microbalances, microbalances, analytical balances, and precision balances excel in speed, accuracy, and quality backed by years of experience.
We offer compact, rugged, and economically priced Sartorius Entris II® and Practum® balances for basic weighing tasks such as laboratory, training, and school use. Models are available with readabilities 1 g to 0.1 mg and weight ranges up to 8.2 kg.
Sartorius Quintix® and Secura® are designed for applications where both accurate weighing results and easy operation are required. They feature Integrated application programs and automatic internal adjustment function. Models are available with readabilities 1 g to 1 μg.
Cubis II balances are the new generation of modular premium lab balances. With a completely configurable high-performance range of software and hardware, unique solutions are available for any application. Models are available with readabilities from 0.1 µg to 1 g.
Sartorius moisture analysers are designed for at-line/on-line use in the laboratory. They provide a non-destructive measurement, determining moisture content without regular wet chemistry—a fast alternative to the oven drying method.