Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Sartorius Cubis II balances are the new generation of modular premium lab balances. With a completely configurable, high-performance range of software and hardware, unique solutions are available for any application. For scientists in R&D or analytical laboratories needing the most reliable lab weighing results, the Cubis II maximises operational efficiency and experimental outcomes. Models are available from 0.1 µg to 1 g.
Sartorius Cubis II microbalances have the highest level of accuracy and precision and can be easily integrated into workflows in a wide range of areas, including QC and analytical labs in the pharma, environmental, and automotive industries, and many more. They feature readability of 0.1 µg or 1 µg, and capacity of up to 10 g. When combined with the optional QApp Pharma Software Package, these balances fulfil requirements for pharma and GxP compliance, including US Pharmacopeia Chapter 41.
With a maximum capacity of between 120 and 520 g and readability between 0.01 and 0.1 mg, Sartorius Cubis II semi-micro and analytical balances provide an excellent solution for maximum weighing performance. With a choice of displays and software packages, Cubis II semi-micro and analytical balances can be configured to suit your specific requirements.
Sartorius Cubis II precision balances are high performing, customisable lab balances for accurate and precise everyday use. Models have a maximum capacity ranging from 320 g to 14,200 g, with a readability range of 1 mg to
100 mg. With many configurations available, these precision balances will provide best-in-class performance regardless of sample type.
Sartorius Cubis II high-capacity balances are designed to weigh between 11,200 and 70,200 g, with readability of 100 mg to 1 g. With customisable, intuitive applications, these high-capacity balances enable faster and more efficient work for enhanced process reliability.
“In 1972, our balances were used for measuring the samples from the moon landing.”
From ultra-micro to high capacity find out how the Sartorius Cubis® II range of laboratory balances can be integrated into any laboratory weighing workflow in any industry.