Sartorius standard balances
Sartorius Quintix® and Secura® are designed for applications where both accurate weighing results and easy operation are required. They feature Integrated application programs and automatic internal adjustment functions. Models are available with readabilities 2 µg to 1 g.
Request information
Featured standard balances
Secura balances
Reduce your risk with integrated and intelligent protection systems. Secura gives you the security and peace of mind of knowing that you have done everything right. Besides providing highly accurate weighing results and operating convenience, Secura also features built-in protection systems for complete reliability and regulatory compliance – the safe and secure way.
Real-time guidance prompts for levelling, automatic internal adjustment, and 100% traceable, clear documentation with sample and batch identifiers make your lab work more efficient.
  • Integrated protection systems for minimising your risk of obtaining incorrect results
  • Real-time alarm messaging and interactive user guidance for levelling to prevent inaccurate results due to an unleveled balance
  • SQmin for automatic detection and active monitoring to ensure you are within the acceptable USP operating range for reliable results
  • Fully automatic internal adjustment function, isoCAL, for the highest possible accuracy at all times
  • Cal Audit Trail for easy, traceable documentation of all calibration and adjustment data
  • Safe and clear documentation by sample and batch identifiers for keeping perfect track of weighing data
  • GLP-|GMP-compliant printouts
  • Standard password security features for protection against tampering and unauthorised access
  • Integrated PC direct function and Plug & Work connectivity to a Sartorius printer
Learn more
Secura specifications
AdjustmentInternal automatic (isoCAL)
Anti-theft locking device
Kensington lock, lockdown capability for cable or chain
Application programs
  • Weighing
  • Density
  • Percentage
  • Checkweighing
  • Peak hold
  • Counting
  • Unstable conditions
  • Animal weighing 
Special built-in lab applications:
  • Mixing 
  • Components 
  • Statistics 
  • Conversion
LevellingReal-time level support
Linearity0.2 - 1000 mg (depending on model)
Operating conditions - temperature+10°C to +30°C
Power requirements100 - 240 VAC, 50 - 60 Hz
Repeatability0.004 mg - 50 mg (depending on model)
Typical stabilisation time≤0.9 s - 6.0 s (depending on model)
Weighing chamber height
209 mm - 219 mm (depending on model)
Weighing unitsGram, kilogram, carat, pound, ounce, troy ounce, Hong Kong tael, Singapore tael, Taiwan tael, grain, pennyweights, milligram, parts per pound, China tael, mommes, Austrian carat, tola, baht, mesghal and Newton
DisplayTouch screen with Sartorius graphical user interface
Maximum weighing capacity21 g - 6100 g (depending on model)
Readability0.002 mg - 100 mg (depending on model)
Weighing pan dimensions⌀ 90 - 180 mm (depending on model)
Data interface
  • mini USB
  • Automatic recognition of Sartorius printer models YDP30 or YDP40
  • PC-direct data transfer to Microsoft® Windows programs
  • Programmable interval for data output
  • Data transfer protocols SBI, xBPI, table format, text format
Quintix balances
Redefining standards with Quintix and makes laboratory work more enjoyable with the Quintix revolutionary user interface. The intuitive touch-screen, in combination with the most precise weighing technology, makes Quintix laboratory balances the only choice to achieve the best results. You will benefit from improved workflows with easy-to-use application programs. If you need to generate GLP-compliant documents, you will save considerable time and effort with the "plug and work" connectivity of Quintix laboratory balances.
  • Intuitive operation of the balance without having to read a manual, thanks to the new user interface and colour touch screen
  • Integrated application programs and intuitive user guidance for greater efficiency in your daily lab routine
  • Highest accuracy and convenience ensured at all times by the isoCAL fully automatic internal adjustment feature
  • Easier record-keeping using the balance's directly integrated PC functions and Plug & Work connectivity to a Sartorius lab printer
Learn more
Quintix specifications
AdjustmentInternal automatic (isoCAL)
Anti-theft locking device
Kensington lock, lockdown capability for cable or chain
Application programs
Standard
  • Weighing
  • Density
  • Percentage
  • Checkweighing
  • Peak hold
  • Counting
  • Unstable conditions
  • Animal weighing
Special built-in lab applications
  • Mixing
  • Components
  • Statistics
  • Conversion
LevellingGlass level indicator with air bubble for centering
Linearity0.2 - 1000 mg (depending on model)
Operating conditions - temperature+10°C to +30°C
Power requirements100 - 240 VAC, 50 - 60 Hz
Repeatability0.02 mg - 500 mg At 5% load, typical value (depending on model)
Typical stabilisation time≤0.9 s - 4.0 s (depending on model)
Weighing chamber height
209 - 219 mm (depending on model)
Weighing unitsGram, kilogram, carat, pound, ounce, troy ounce, Hong Kong tael, Singapore tael, Taiwan tael, grain, pennyweights, milligram, parts per pound, China tael, mommes, Austrian carat, tola, baht, mesghal and Newton
Display
Touch screen with Sartorius graphical user interface
Maximum weighing capacity30 g - 6100 g (depending on model)
Readability0.01 mg - 1000 mg (depending on model)
Weighing pan dimensions⌀ 90 - 180 mm (depending on model)
Data interface
  • Mini USB
  • Automatic recognition of Sartorius - printer models YDP30 or YDP40
  • PC-direct data transfer to Microsoft® Windows programs
  • Programmable interval for data output
  • Data transfer protocols SBI, xBPI, table format, text format
Secura balances
Reduce your risk with integrated and intelligent protection systems. Secura gives you the security and peace of mind of knowing that you have done everything right. Besides providing highly accurate weighing results and operating convenience, Secura also features built-in protection systems for complete reliability and regulatory compliance – the safe and secure way.
Real-time guidance prompts for levelling, automatic internal adjustment, and 100% traceable, clear documentation with sample and batch identifiers make your lab work more efficient.
  • Integrated protection systems for minimising your risk of obtaining incorrect results
  • Real-time alarm messaging and interactive user guidance for levelling to prevent inaccurate results due to an unleveled balance
  • SQmin for automatic detection and active monitoring to ensure you are within the acceptable USP operating range for reliable results
  • Fully automatic internal adjustment function, isoCAL, for the highest possible accuracy at all times
  • Cal Audit Trail for easy, traceable documentation of all calibration and adjustment data
  • Safe and clear documentation by sample and batch identifiers for keeping perfect track of weighing data
  • GLP-|GMP-compliant printouts
  • Standard password security features for protection against tampering and unauthorised access
  • Integrated PC direct function and Plug & Work connectivity to a Sartorius printer
Learn more
Secura specifications
AdjustmentInternal automatic (isoCAL)
Anti-theft locking device
Kensington lock, lockdown capability for cable or chain
Application programs
  • Weighing
  • Density
  • Percentage
  • Checkweighing
  • Peak hold
  • Counting
  • Unstable conditions
  • Animal weighing 
Special built-in lab applications:
  • Mixing 
  • Components 
  • Statistics 
  • Conversion
LevellingReal-time level support
Linearity0.2 - 1000 mg (depending on model)
Operating conditions - temperature+10°C to +30°C
Power requirements100 - 240 VAC, 50 - 60 Hz
Repeatability0.004 mg - 50 mg (depending on model)
Typical stabilisation time≤0.9 s - 6.0 s (depending on model)
Weighing chamber height
209 mm - 219 mm (depending on model)
Weighing unitsGram, kilogram, carat, pound, ounce, troy ounce, Hong Kong tael, Singapore tael, Taiwan tael, grain, pennyweights, milligram, parts per pound, China tael, mommes, Austrian carat, tola, baht, mesghal and Newton
DisplayTouch screen with Sartorius graphical user interface
Maximum weighing capacity21 g - 6100 g (depending on model)
Readability0.002 mg - 100 mg (depending on model)
Weighing pan dimensions⌀ 90 - 180 mm (depending on model)
Data interface
  • mini USB
  • Automatic recognition of Sartorius printer models YDP30 or YDP40
  • PC-direct data transfer to Microsoft® Windows programs
  • Programmable interval for data output
  • Data transfer protocols SBI, xBPI, table format, text format
Quintix balances
Redefining standards with Quintix and makes laboratory work more enjoyable with the Quintix revolutionary user interface. The intuitive touch-screen, in combination with the most precise weighing technology, makes Quintix laboratory balances the only choice to achieve the best results. You will benefit from improved workflows with easy-to-use application programs. If you need to generate GLP-compliant documents, you will save considerable time and effort with the "plug and work" connectivity of Quintix laboratory balances.
  • Intuitive operation of the balance without having to read a manual, thanks to the new user interface and colour touch screen
  • Integrated application programs and intuitive user guidance for greater efficiency in your daily lab routine
  • Highest accuracy and convenience ensured at all times by the isoCAL fully automatic internal adjustment feature
  • Easier record-keeping using the balance's directly integrated PC functions and Plug & Work connectivity to a Sartorius lab printer
Learn more
Quintix specifications
AdjustmentInternal automatic (isoCAL)
Anti-theft locking device
Kensington lock, lockdown capability for cable or chain
Application programs
Standard
  • Weighing
  • Density
  • Percentage
  • Checkweighing
  • Peak hold
  • Counting
  • Unstable conditions
  • Animal weighing
Special built-in lab applications
  • Mixing
  • Components
  • Statistics
  • Conversion
LevellingGlass level indicator with air bubble for centering
Linearity0.2 - 1000 mg (depending on model)
Operating conditions - temperature+10°C to +30°C
Power requirements100 - 240 VAC, 50 - 60 Hz
Repeatability0.02 mg - 500 mg At 5% load, typical value (depending on model)
Typical stabilisation time≤0.9 s - 4.0 s (depending on model)
Weighing chamber height
209 - 219 mm (depending on model)
Weighing unitsGram, kilogram, carat, pound, ounce, troy ounce, Hong Kong tael, Singapore tael, Taiwan tael, grain, pennyweights, milligram, parts per pound, China tael, mommes, Austrian carat, tola, baht, mesghal and Newton
Display
Touch screen with Sartorius graphical user interface
Maximum weighing capacity30 g - 6100 g (depending on model)
Readability0.01 mg - 1000 mg (depending on model)
Weighing pan dimensions⌀ 90 - 180 mm (depending on model)
Data interface
  • Mini USB
  • Automatic recognition of Sartorius - printer models YDP30 or YDP40
  • PC-direct data transfer to Microsoft® Windows programs
  • Programmable interval for data output
  • Data transfer protocols SBI, xBPI, table format, text format