Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
We offer a complete range of Labconco freeze dryers to handle the lyophilisation requirements of research and pilot plant labs. Sizes are available from 2.5 L to
18 L, with a choice of collector temperatures to suit different chemical applications. A large selection of drying accessories are also available, including flasks, bulk drying chambers, and stoppering tray dryers.
Benchtop | Console | Triad benchtop |
Suitable for lyophilising light to moderate sample loads, these benchtop freeze dryers have a small footprint and are ideal for labs with limited floor space. Combine with the Lyo-Works™ OS for remote notification and data collection. | Console freeze dryers are suitable for moderate to large sample loads or numerous sample batches. Combine with a wide range of accessories to customise the dryer for any application. These console freeze dryers can also be combined with the stoppering tray dryer in one easy-to-order code, or with the bulk tray dryer as an additional, separately ordered, accessory. | FreeZone Triad benchtop freeze dryers combine a benchtop lyophiliser with a built-in stoppering tray dryer for vials and four-port manifold for flasks. All that is required is a vacuum pump, and you’re ready to go. |
2.5, 4.5, or 8 L capacity | 6, 12, or 18 L capacity | 2.5 L capacity |
–50, –84, or –105°C | –50 or –84°C | –84°C |
FreeZone Benchtop Freeze Dryers › FreeZone Legacy Freeze Dryers › Dry Ice Benchtop Freeze Dryers › | FreeZone Console Freeze Dryers › | FreeZone Triad Benchtop Freeze Dryers › |
Labconco FreeZone tray dryers are the ideal drying accessory for freeze dryers, whether you are stoppering vials or have numerous samples batches. Confidently pre-freeze and lyophilise samples with the precise shelf temperature control. Stoppering tray dryers can be supplied with console models in one easy-to-order code or ordered separately. Tray dryers are to be used with freeze dryers with Lyo-Works OS only.
Combine your FreeZone freeze dryer with a wide range of drying accessories to create a custom solution for your specific application.
Gail's story changed the way we think about laboratory freeze drying
When Gail visited Labconco, her story led to fundamental changes in how laboratory freeze drying is done. With the creation of the Lyo-Works Operating System, samples can now be monitored from anywhere via smartphone. And with End-Zone™ End Point Detection, FreeZone Freeze Dryers can even send you an alert when your samples have lyophilized completely.