Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Finntip and Finntip Flex tips are engineered to make life in your lab safer, easier, and more successful. Designed and manufactured expressly for manual and electronic Finnpipettes, these tips are part of a Finnpipette system that has become a trusted standard of excellence for comfort, ease-of-use, and reliability.
These tips are engineered for a secure seal you can feel with your Finnpipettes, creating a pipetting system that delivers optimal accuracy and precision.
Finntip Flex tips are designed with a softer, flexible collar for an ergonomic fit with your Finnpipette to maximize performance and comfort, with reduced attachment and ejection forces to minimize hand strain.
Pipetting styles vary, so pair your Finnpipette with a compatible tip to achieve the system that performs best for your pipetting technique, with optimal precision and accuracy with every combination.
Below are some suggested systems, although you can pair all Finntip and Finntip Flex tips with any Finnpipette.
Personalize your Finnpipette pipetting system | |||
Your need | Finnpipette | Suggested tip | Performance |
Enhanced ergonomics When you pipette all day and need to minimize hand strain |
| Finntip Flex |
|
Durability For use in demanding applications and environments |
| Finntip |
|
Higher throughput For productivity and higher throughput needs | Finntip Flex |
|
Tip type | Benefits | Recommended use |
Filtered tip |
|
|
Non-filtered tip |
|
|
Extended-length tip |
|
|
Micropoint tip |
|
|
Macro tip |
|
|
Wide-bore tip |
|
|
Finntip racks and Finnpipettes are color coded by volume, making it easy to select compatible tips at a glance.
Color | Volume range |
Pink | 10 µL |
Purple | 20 µL |
Turquoise | 20 µL / 50 µL |
Yellow | 100 µL / 200 µL |
Orange | 300 µL |
Blue | 1000 µL |
Green | 5 mL |
Red | 10 mL |
Packaging type | Features | |
Finntip hinged rack |
| |
Finntip Flex hinged rack |
| |
Finntip reload kit |
| |
Finntip Flex reload kit |
| |
Bulk |
| |
Individually wrapped |
|
Tip type | Benefits | Recommended use |
Filtered tip |
|
|
Non-filtered tip |
|
|
Extended-length tip |
|
|
Micropoint tip |
|
|
Macro tip |
|
|
Wide-bore tip |
|
|
Finntip racks and Finnpipettes are color coded by volume, making it easy to select compatible tips at a glance.
Color | Volume range |
Pink | 10 µL |
Purple | 20 µL |
Turquoise | 20 µL / 50 µL |
Yellow | 100 µL / 200 µL |
Orange | 300 µL |
Blue | 1000 µL |
Green | 5 mL |
Red | 10 mL |
Packaging type | Features | |
Finntip hinged rack |
| |
Finntip Flex hinged rack |
| |
Finntip reload kit |
| |
Finntip Flex reload kit |
| |
Bulk |
| |
Individually wrapped |
|
Filtered, non-filtered or specialty pipette tips: Finntip and Finntip Flex portfolios offer a variety of filtered tip options for any application.
Filtered tips | Non-filtered tips | Specialty tips |
Filters protect samples and pipettes by blocking aerosols and contaminants. Ideal for use in the following applications:
| Non-filtered Finntip and Finntip Flex tips are available in sterile and non-sterile options for use in:
| Specialty tips are designed for unique laboratory applications. Whether pipetting fragile cell lines, transferring liquids from tall vessels, or extra accuracy is required for large volumes, there is a Finntip product for you.
Specialty tips include:
|
Use Finntip and and Finntip Flex tips with any pipette in your lab
Researchers worldwide love the fit and performance of Finntip and Finntip Flex so much that they often use them with all of their pipettes. You can too. Consider choosing our Finntip 250 Universal pipette tip and any Finntip Flex tip for a secure fit and accurate, precise pipetting.