Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Thermo Scientific SoftFit-L pipette tips are designed to fit Rainin LTS style pipettes. The cylindrical collar sharp and positive – stop design ensure a perfect fit, reducing attachment and ejection forces. SoftFit-L tips provide the accuracy, precision, and comfort you have come to expect when using your Rainin LTS pipette.
Hydrophobic surface of low retention tips
Color-coded insert for matching tip and pipette volume
Certified to be free of Rnase, Dnase, DNA, ATP and Endotoxins
Tip type | Features | Applications |
SoftFit-L filtered tip* | Blocks aerosols from contaminating the pipette and prevent cross-contamination of samples |
|
SoftFit-L non-filtered tip | Economic, high-quality tips |
|
SoftFit-L low retention tip | Hydrophobic surface maximizes sample delivery and conservation of expensive reagents |
|
Package type | Features |
Hinged Rack |
|
Reload insert* |
|
Reload tower* |
|
*SoftFit-L reload towers and inserts are compatible with SoftFit-L tip racks only. Not for use with other manufacturer’s racks. SoftFit-L racks can be ordered separately.
SoftFit-L Empty Racks for use with SoftFit-L Reload Inserts and Reload System Towers.
Tip type | Features | Applications |
SoftFit-L filtered tip* | Blocks aerosols from contaminating the pipette and prevent cross-contamination of samples |
|
SoftFit-L non-filtered tip | Economic, high-quality tips |
|
SoftFit-L low retention tip | Hydrophobic surface maximizes sample delivery and conservation of expensive reagents |
|
Package type | Features |
Hinged Rack |
|
Reload insert* |
|
Reload tower* |
|
*SoftFit-L reload towers and inserts are compatible with SoftFit-L tip racks only. Not for use with other manufacturer’s racks. SoftFit-L racks can be ordered separately.
SoftFit-L Empty Racks for use with SoftFit-L Reload Inserts and Reload System Towers.