Search Thermo Fisher Scientific
Catalog # | Anti-fluorophore | Host | Label |
---|---|---|---|
Anti-fluorophore antibodies and their conjugates | |||
A5760 | Alexa Fluor® 405/Cascade Blue® dye | Rabbit | None |
A11094 | Alexa Fluor® 488 dye | Rabbit | None |
A5770 | BODIPY® FL dye | Rabbit | None |
A6398 | Dansyl | Rabbit | None |
MA514696 | Fluorescein (FITC) | Mouse (clone #9) | None |
A6421 | Fluorescein/Oregon Green® dye | Mouse (clone 4-4-20) | None |
A889 | Fluorescein/Oregon Green® dye | Rabbit | None |
A6413 | Fluorescein/Oregon Green® dye | Rabbit, Fab fragment | None |
A11095 | Fluorescein/Oregon Green® dye | Goat | None |
A11090 | Fluorescein/Oregon Green® dye | Rabbit | Alexa Fluor® 488 dye |
A11096 | Fluorescein/Oregon Green® dye | Goat | Alexa Fluor® 488 dye |
A11091 | Fluorescein/Oregon Green® dye | Rabbit | Alexa Fluor® 594 dye |
A21250 | Fluorescein/Oregon Green® dye | Rabbit | R-phycoerythrin |
A982 | Fluorescein/Oregon Green® dye | Rabbit | Biotin-XX |
A21253 | Fluorescein/Oregon Green® dye | Rabbit | Horseradish peroxidase |
A5750 | Lucifer yellow dye | Rabbit | None |
A5751 | Lucifer yellow dye | Rabbit | Biotin-XX |
MA1041 | Tetramethylrhodamine (TAMRA) | Mouse (clone 5G5) | None |
A6397 | Tetramethylrhodamine (TAMRA) and Rhodamine Red™ dye | Rabbit | None |
A6399 | Texas Red® and Texas Red®-X dye | Rabbit | None |
Anti-hapten antibodies and their conjugates | |||
MA511251 | Biotin | Mouse (clone BTN.4) | None |
033700 | Biotin | Mouse (clone Z021) | None |
A6430 | Dinitrophenyl | Rabbit | None |
A6435 | Dinitrophenyl | Rabbit | Biotin-XX |
A11097 | Dinitrophenyl | Rabbit | Alexa Fluor® 488 |
A6423 | Dinitrophenyl | Rabbit | Fluorescein |
MA135779 | Dinitrophenyl | Rat (clone LO-DNP-2) | None |
A21285 | Nitrotyrosine | Rabbit | None |
MA112770 | 3-Nitrotyrosine | Mouse (clone 2A12) | None |
Partner with us to scale-up your process or product development pipeline with bulk quantities of high-quality immunoassay reagents and biochemicals. We offer a variety of compounds, devices, and resins for your large-scale applications.