Search Thermo Fisher Scientific
Protease and phosphatase inhibitor cocktails, tablets, and capsules are ideal for the protection of proteins during extraction or lysate preparation from primary cells, cultured mammalian cells, animal tissues, plant tissues, yeast, or bacterial cells. Individual protease inhibitors are also available separately and in multiple sizes. Formulations with or without EDTA are available for divalent cation-sensitive assays. Find the right protease, phosphatase, or combined liquid cocktails, tablets, and capsules for your experiments below.
Liquid | Liquid, EDTA-free | Tablet | Tablet, EDTA-free | |
---|---|---|---|---|
Halt Protease Inhibitor Cocktail | Halt Protease Inhibitor Cocktail, EDTA-free | |||
Contains EDTA | Yes, but in a separate vial | No | Yes | No |
Contains DMSO | Yes | Yes | No | No |
Requires reconstitution | No (100X concentrate) | No (100X concentrate) | Yes | Yes |
Downstream compatibility | Not compatible with 2D or IMAC | All | Not compatible with 2D or IMAC | All |
Included inhibitors | AEBSF, Aprotinin, Bestatin, E-64, Leupeptin, Pepstatin A, EDTA* | |||
Cat. No. | 78430 (24 x 100 µL) 78429 (5 mL) 78438 (10 mL) 87786 (1 mL) | 78425 (24 x 100 µL) 78437 (5 mL) 78439 (10 mL) 87785 (1 mL) | A32963 (50 mL buffer per tablet) A32953 (Mini, 10 mL buffer per tablet) | A32965 (50 mL buffer per tablet) A32955 (Mini, 10 mL buffer per tablet) A37989 (XL, 500 mL buffer per capsule) |
*Only in EDTA formulations
Figure 1. Pierce Protease Inhibitor tablets inhibits protease activity in different tissue lysates and cell lines. Protease activity of cell and tissue lysates (1 mg/mL) was determined in the presence (blue bars) and absence (black bars) of prepared Pierce Protease Inhibitor Tablet. The percentage of protease inhibition is indicated.
Liquid | Tablet | |
---|---|---|
Halt Phosphatase Inhibitor Cocktail | Pierce Phosphatase Inhibitor Tablet | |
Contains DMSO | Yes | No |
Requires reconstitution | No (100X concentrate) | Yes |
Downstream compatibility | Not compatible with 2D or IMAC | All |
Included inhibitors | Sodium fluoride, sodium orthovanadate, sodium pyrophosphate, β-glycerophosphate | |
Cat. No. | 78428 (24 x 100 uL) 78420 (1 mL) 78426 (5 x 1 mL) 78427 (10 mL) | A32957 (Mini, 10 mL buffer per tablet) |
Liquid | Liquid, EDTA-free | Tablet | Tablet, EDTA-free | |
---|---|---|---|---|
Halt Protease and Phosphatase Inhibitor Cocktail | Halt Protease and Phosphatase Inhibitor Cocktail, EDTA-free | Pierce Protease and Phosphatase Inhibitor Tablets | Pierce Protease and Phosphatase Inhibitor Tablets, EDTA-free | |
Contains EDTA | Yes, but in a separate vial | No | Yes | No |
Included inhibitors | Aprotinin, bestatin, E-64, leupeptin, sodium fluoride, sodium orthovanadate, sodium pyrophosphate, β-glycerophosphate, EDTA* | |||
Contains DMSO | Yes | Yes | No | No |
Requires reconstitution | No (100X concentrate) | No (100X concentrate) | Yes | Yes |
Downstream compatibility | Not compatible with 2D or IMAC | All | Not compatible with 2D or IMAC | All |
Cat. No. | 78442 (24 x 100 μL) 78440 (1 mL) 78444 (5 x 1 mL) 78446 (10 mL) | 78443 (24 x 100 μL) 78441 (1 mL) 78445 (5 x 1 mL) 78447 (10 mL) | A32959 (Mini, 10 mL buffer per tablet) | A32961 (Mini, 10 mL buffer per tablet) |
*Only in EDTA formulations
These combined protease and phosphatase inhibitor cocktails and tablets contain chemical compounds that target and inhibit serine, threonine, and tyrosine phosphatases, as well as serine, cysteine, and aspartic acid proteases, as well as aminopeptidases. Metalloproteases are inhibited by the addition of EDTA, which is available in a separate vial in the liquid format but included in the tablet format. These broad-spectrum inhibitor formulations prevent protein degradation and preserve phosphorylation simultaneously, helping provide complete protection in a single solution or tablet.
Protease Inhibitors
Phosphatase Inhibitors
Combined Protease and Phosphatase Inhibitors
Liquid | Liquid, EDTA-free | Tablet | Tablet, EDTA-free | |
---|---|---|---|---|
Halt Protease Inhibitor Cocktail | Halt Protease Inhibitor Cocktail, EDTA-free | |||
Contains EDTA | Yes, but in a separate vial | No | Yes | No |
Contains DMSO | Yes | Yes | No | No |
Requires reconstitution | No (100X concentrate) | No (100X concentrate) | Yes | Yes |
Downstream compatibility | Not compatible with 2D or IMAC | All | Not compatible with 2D or IMAC | All |
Included inhibitors | AEBSF, Aprotinin, Bestatin, E-64, Leupeptin, Pepstatin A, EDTA* | |||
Cat. No. | 78430 (24 x 100 µL) 78429 (5 mL) 78438 (10 mL) 87786 (1 mL) | 78425 (24 x 100 µL) 78437 (5 mL) 78439 (10 mL) 87785 (1 mL) | A32963 (50 mL buffer per tablet) A32953 (Mini, 10 mL buffer per tablet) | A32965 (50 mL buffer per tablet) A32955 (Mini, 10 mL buffer per tablet) A37989 (XL, 500 mL buffer per capsule) |
*Only in EDTA formulations
Figure 1. Pierce Protease Inhibitor tablets inhibits protease activity in different tissue lysates and cell lines. Protease activity of cell and tissue lysates (1 mg/mL) was determined in the presence (blue bars) and absence (black bars) of prepared Pierce Protease Inhibitor Tablet. The percentage of protease inhibition is indicated.
Liquid | Tablet | |
---|---|---|
Halt Phosphatase Inhibitor Cocktail | Pierce Phosphatase Inhibitor Tablet | |
Contains DMSO | Yes | No |
Requires reconstitution | No (100X concentrate) | Yes |
Downstream compatibility | Not compatible with 2D or IMAC | All |
Included inhibitors | Sodium fluoride, sodium orthovanadate, sodium pyrophosphate, β-glycerophosphate | |
Cat. No. | 78428 (24 x 100 uL) 78420 (1 mL) 78426 (5 x 1 mL) 78427 (10 mL) | A32957 (Mini, 10 mL buffer per tablet) |
Liquid | Liquid, EDTA-free | Tablet | Tablet, EDTA-free | |
---|---|---|---|---|
Halt Protease and Phosphatase Inhibitor Cocktail | Halt Protease and Phosphatase Inhibitor Cocktail, EDTA-free | Pierce Protease and Phosphatase Inhibitor Tablets | Pierce Protease and Phosphatase Inhibitor Tablets, EDTA-free | |
Contains EDTA | Yes, but in a separate vial | No | Yes | No |
Included inhibitors | Aprotinin, bestatin, E-64, leupeptin, sodium fluoride, sodium orthovanadate, sodium pyrophosphate, β-glycerophosphate, EDTA* | |||
Contains DMSO | Yes | Yes | No | No |
Requires reconstitution | No (100X concentrate) | No (100X concentrate) | Yes | Yes |
Downstream compatibility | Not compatible with 2D or IMAC | All | Not compatible with 2D or IMAC | All |
Cat. No. | 78442 (24 x 100 μL) 78440 (1 mL) 78444 (5 x 1 mL) 78446 (10 mL) | 78443 (24 x 100 μL) 78441 (1 mL) 78445 (5 x 1 mL) 78447 (10 mL) | A32959 (Mini, 10 mL buffer per tablet) | A32961 (Mini, 10 mL buffer per tablet) |
*Only in EDTA formulations
These combined protease and phosphatase inhibitor cocktails and tablets contain chemical compounds that target and inhibit serine, threonine, and tyrosine phosphatases, as well as serine, cysteine, and aspartic acid proteases, as well as aminopeptidases. Metalloproteases are inhibited by the addition of EDTA, which is available in a separate vial in the liquid format but included in the tablet format. These broad-spectrum inhibitor formulations prevent protein degradation and preserve phosphorylation simultaneously, helping provide complete protection in a single solution or tablet.
Protease Inhibitors
Phosphatase Inhibitors
Combined Protease and Phosphatase Inhibitors