Search Thermo Fisher Scientific
Discover our modern, powerful and robust clinical prenatal screening software for pre-eclampsia in first trimester.
Thermo Scientific B·R·A·H·M·S Fast Screen pre I plus is a CE marked prenatal screening software. This software, designed to be used with Thermo Scientific B·R·A·H·M·S KRYPTOR Instruments and Assays, ensures convenience of data entry, risk calculation and reporting for laboratories with both low and high data throughput.
B·R·A·H·M·S Fast Screen pre I plus comprises algorithms that are based on the Fetal Medicine Foundation UK (FMF) data and are updated with recently published correction factors.
Pre-eclampsia | |
Time range |
|
Biomarkers |
|
Physical markers |
|
The medians of the B·R·A·H·M·S biochemical markers free βhCG, PAPP-A and PlGF which are included in our software are extremely stable. These medians were established on over 220,000 healthy pregnancies which ensures reproducibility and robustness of the risk calculation.1,2 Therefore, a recalculation of the medians and MoMs over time is not required.
View the FMF pre-eclampsia algorithm performances below.3
Early onset pre-eclampsia (< week 32) |
Intermediate onset pre-eclampsia (< week 37) |
Late onset pre-eclampsia (≥ week 37) |
|
Detection rate | 100% | 75% | 43% |
False positive rate | 10% | 10% | 10% |
Simple and user-friendly interface available in six languages
Available in English, German, French, Spanish, Italian, and Turkish
Thermo Fisher Scientific products are distributed globally and their uses, applications, indications, claims and availability of products in each country depend on local regulatory marketing authorization status, please consult the Instructions For Use (IFU) available in your country.
© 2024 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified. B·R·A·H·M·S is a registered trademark of B·R·A·H·M·S GmbH.
KRYPTOR and TRACE are trademarks of Cisbio Bioassays, licensed for use by B·R·A·H·M·S GmbH, a part of Thermo Fisher Scientific. Nobel Prize is a registered trademark of the Nobel Foundation.