Search Thermo Fisher Scientific
SYBR GreenER qPCR SuperMix for ABI PRISM is a ready-to-use cocktail containing all components, except primers and template, for real-time quantitative PCR (qPCR) on ABI real-time instruments that support normalization with ROX Reference Dye at a final concentration of 500 nM. It combines a chemically modified “hot-start” version of Taq DNA polymerase with integrated uracil DNA glycosylase (UDG) carryover prevention technology and a novel fluorescent dye to deliver excellent sensitivity in the quantification of target sequences, with a linear dose response over a wide range of target concentrations.
Volumes are provided for 100 or 500 amplification reactions of 50 μl each.
SYBR GreenER qPCR SuperMix for ABI PRISM is supplied at a 2X concentration and contains hot-start Taq DNA polymerase, SYBR GreenER fluorescent dye, 1 μM ROX Reference Dye, MgCl2, dNTPs (with dUTP instead of dTTP), UDG, and stabilizers. The SuperMix formulation can quantify fewer than 10 copies of a target gene, has a broad dynamic range, and is compatible with melting curve analysis.
Component | 100-rxn Kit | 500-rxn Kit |
SYBR GreenER qPCR SuperMix for ABI PRISM | 2 × 1.25 ml | 12.5 ml |
The SuperMix is shipped on dry ice and should be stored at 4ºC. Storage at –20ºC may extend shelf life.
Minimize exposure of SYBR GreenER qPCR SuperMix for ABI PRISM to direct light. Exposure to direct light for an extended period of time may result in loss of fluorescent signal intensity.
This SuperMix is also included with the SYBR GreenER Two-Step qRT-PCR Kit for ABI PRISM, catalog nos. 11763-100 and 11763-500.
The Certificate of Analysis (CofA) provides detailed quality control information for each product. Search for the CofA for your product.
This kit can be used with ABI real-time instruments that are compatible with ROX Reference Dye at a final concentration of 500 nM. These instruments include the ABI PRISM 7000, 7700, and 7900HT; the ABI 7300 Real-Time PCR System; and the ABI GeneAmp 5700.
Note: This kit is not compatible with instruments that use ROX at a final concentration lower than 500 nM, including the ABI 7500. For these instruments, we recommend SYBR GreenER qPCR SuperMix Universal, which includes ROX as a separate tube that can be added at the required concentration (see Additional Products in Ordering Table).
Use up to 100 ng of genomic DNA or 10–107 copies of plasmid DNA in a 10-μl volume. Note that 1 μg of plasmid DNA contains 9.1 × 1011 copies divided by the plasmid size in kilobases.
Primer design is one of the most important parameters when using SYBR GreenER qPCR SuperMix. We strongly recommend using a primer design program such as OligoPerfect, available here, or Vector NTI. When designing primers, the amplicon length should be approximately 80–250 bp. A final concentration of 200 nM per primer is effective for most reactions. Optimal results may require a titration of primer concentrations between 100 and 500 nM.
The hot-start DNA polymerase is activated in the 10-minute incubation at 95°C before PCR cycling.
Melting curve analysis should always be performed following real-time qPCR to identify the presence of primer dimers and analyze the specificity of the reaction. Program your instrument for melting curve analysis using the instructions provided with your specific instrument.
Magnesium chloride is included in the SuperMix at an optimized concentration for qPCR.
Component | Single rxn | Notes |
---|---|---|
SYBR® GreenER™ qPCR SuperMix for ABI PRISM® | 25 μl | 1X final conc. |
Forward primer, 10 μM | 1 μl | 200 nM final conc. |
Reverse primer, 10 μM | 1 μl | 200 nM final conc |
Template (up to 100 ng of genomic DNA, 10–107 copies of plasmid DNA, or cDNA generated from up to 1 μg of total RNA) | 5–10 μl | max. 10% v/v undiluted cDNA |
DEPC-treated water | to 50 μl | — |
Problem | Possible Cause | Solution |
---|---|---|
Signals are present in no-template controls, and/or multiple peaks are present in the melting curve graph | Template or reagents are contaminated by nucleic acids (DNA, cDNA) | Use melting curve analysis and/or run the PCR products on a 4% agarose gel after the reaction to identify contaminants. Take standard precautions to avoid contamination when preparing your PCR reactions. Ideally, amplification reactions should be assembled in a DNA-free environment. We recommend using aerosol-resistant barrier tips. |
Primer dimers or other primer artifacts are present | Use melting curve analysis to identify primer dimers. We recommend using validated pre-designed primer sets or design primers using dedicated software programs or primer databases. Primer contamination or truncated or degraded primers can lead to artifacts. Check the purity of your primers by gel electrophoresis. | |
No amplification curve appears on the qPCR graph | There is no PCR product | Run the reaction on a gel to determine whether PCR worked. Then proceed to the troubleshooting steps below. |
No PCR product is evident, either in the qPCR graph or on a gel | The protocol was not followed correctly | Verify that all steps have been followed and the correct reagents, dilutions, volumes, and cycling parameters have been used. |
Template contains inhibitors, nucleases, or proteases, or has otherwise been degraded. | Purify or re-purify your template. | |
Primer design is suboptimal | Verify your primer selection. We recommend using validated pre-designed primers or design primers using dedicated software programs or primer databases. | |
PCR product is evident in the gel, but not on the qPCR graph | qPCR instrument settings are incorrect | Confirm that you are using the correct instrument settings (dye selection and calibration, reference dye, filters, acquisition points, etc.). |
Problems with your specific qPCR instrument | See your instrument manual for tips and troubleshooting. | |
PCR efficiency is above 110% | Template contains inhibitors, nucleases, or proteases, or has otherwise been degraded. | Purify or re-purify your template. Inhibitors in the template may result in changes in PCR efficiency between dilutions |
Nonspecific products may be amplified. | Use melting curve analysis if possible, and/or run the PCR products on a 4% agarose gel after the reaction to identify contaminants. Suboptimal primer design may lead to nonspecific products. Use validated pre-designed primers or design primers using dedicated software programs or primer databases. | |
PCR efficiency is below 90% | The PCR conditions are suboptimal | Verify that the reagents you are using have not been freeze-thawed multiple times and have not remained at room temperature for too long. Verify that the amount of primers you are using is correct. |