A comprehensive next-generation sequencing (NGS) assay for translational and early clinical research
A comprehensive next-generation sequencing (NGS) assay for translational and early clinical research
The Oncomine Comprehensive Assay v3 is a multi-biomarker NGS assay that covers 161 of the most relevant cancer genes, based on the Oncomine Knowledgebase and from collaborations with leading cancer research institutions and pharmaceutical companies. The low sample input requirement enables the analysis of even small and challenging formalin-fixed paraffin-embedded (FFPE) samples.
Order now  Download flyer  Learn more
Highlights of the Oncomine Comprehensive Assay v3 include:
  • Only 10 ng of DNA or RNA sample input, making the Oncomine Comprehensive Assay v3 an ideal solution for the analysis of challenging FFPE samples
  • Detects relevant SNVs, CNVs, gene fusions, and indels from 161 unique cancer-associated genes in one streamlined workflow
  • High specificity, sensitivity, and reproducibility across different research laboratories, as demonstrated in translational and clinical research projects including the NCI Match Trial
  • Efficient workflow on Ion Chef System plus Ion GeneStudio S5 or Ion Torrent Genexus system with high multiplexing flexibility and fast time to results
"The requirement of a lower DNA input for the Oncomine assay is a significant advantage when primary samples are becoming increasingly limited."
—John Bartlett, PhD
Director of Transformative Pathology Platform
Ontario Institute for Cancer Research
Oncomine Comprehensive Assay v3 gene content
The Oncomine Comprehensive Assay v3 includes:
  • Hotspot SNVs and indels, CNVs, and gene fusions
  • Kinase domain coverage to span the entire kinase domain in several critical receptor tyrosine kinases. This increases the likelihood of detecting functionally relevant mutations in addition to known actionable and prevalent variants (ALK, BRAF, DDR2, EGFR, ERBB2, KIT, MET, PDGFRA, RET, ROS1).
  • Representation of genes involved in DNA repair (31 genes in DNA repair pathway)
  • Coverage of MAPK, PI3K, and cell cycle pathway genes
161 genes categorized by somatic alteration type
87 Hotspots genes
43 Focal CNV gains
48 Full CDS for DEL mutations
51 Fusion drivers
To see the full gene content of the Oncomine Comprehensive Assay v3, download the flyer.
Oncomine Comprehensive Assay v3 performance on Ion GeneStudio S5 sequencer
True positive
False positive
False negative
Sensitivity
Positive predictive value
Hotspots
504
9
4
99.21%
98.25%
Indels
31
0
1
96.88%
100%
DeNovo SNV
602
11
18
97.10%
98.21%
De novo InDel
24
2
4
85.71%
92.31%
Fusions
82
4
4
95.35%
95.35%
Table 1. The performance of the Oncomine Comprehensive Assay v3 has been verified using 20 FFPE cancer tissue samples and commercially available standard material from HorizonDx and SeraCare.
Assay analytical validation—results of the NCI Match Trial
Assay analytical validation--results of the NCI Match Trial
Data adapted from: Lih CJ, et al. Analytical validation of the NGS assay for a nationwide signal-finding clinical trial molecular analysis for therapy choice clinical trial. J Mol Diagn. 2017 Mar;19(2):313-327.
Flexible configurations to fit your needs
The Oncomine Comprehensive Assay v3 has been optimized for throughput of eight samples on the Ion GeneStudio S5 Systems or six samples on the Ion Torrent Genexus System. The assay is available on Ion GeneStudio S5 Systems in two configurations: manual and automated (using Ion Chef System for library preparation) and includes the DNA and RNA primer panel and library preparation reagents. The Genexus System combines preparation, templating, and sequencing in one system.
Ordering information