Search Thermo Fisher Scientific
Clone PCR-amplified DNA fragments (blunt or A-overhang) directly into a choice of over 40 subcloning, sequencing, or expression vectors in just 5 minutes—and obtain up to 95% recombinant clones.
Figure 1. The TOPO cloning protocol. TOPO PCR cloning requires just three easy steps. Simply combine your PCR product and a TOPO cloning vector in the provided solution, wait five minutes, then transform E. coli.
Steps | TOPO TA Cloning Kits | TA/UA Cloning | Restriction enzyme or cut-back cloning |
---|---|---|---|
Use of existing primers? | Yes | Yes | No |
Vector is ready for cloning? | Yes | Yes | No |
Ligation reagents included? | Yes | Yes | No |
Prepare or purchase competent cells separately? | No, included | Purchase: 0 hr Prepare: up to 6 hr | Purchase: 0 hr Prepare: up to 6 hr |
Ligation time | 5 min | 1 hr | 2–23 hr |
Recombination efficiency | Up to 95% | 60–80% | ~60% |
Time required for cloning | 5 min | 1–12 hr | 2–23 hr |