Search Thermo Fisher Scientific
Molecular Probes® fluorescent nucleic acid gel stains—SYBR® Gold, SYBR® Green I, SYBR® Green II, and SYBR® Safe dyes—are highly sensitive reagents for staining RNA in electrophoresis gels. These gel stains provide greater sensitivity, with lower background fluorescence, than the conventional ethidium bromide gel stain.
Detection of nucleic acids in agarose gels & CsCl gradients | Safer, smarter alternative to EtBr | Sensitive stain for any gel system | Ultimate sensitivity for RNA & ssDNA | |
---|---|---|---|---|
Top Seller | ![]() | |||
Sensitivity - RNA | Sensitive (1.5 ng) | Sensitive (1.5 ng) | Sensitive (1.5 ng) | Ultra Sensitive (100 pg) |
Lowest Background | ![]() | |||
Nonhazardous/ Environmentally-Friendly | ![]() | |||
Available as Ready to Use | ![]() | |||
Ideal for Multiple Applications | ![]() | |||
Package sizes | 10 mL | 0.4 mL, 1 L (ready to use), & 4 L (ready to use) | 0.5 mL | 0.5 mL & 1 mL |
Order Now | Order Now | Order Now | Order Now |